Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RIMS1 SIRNA Assays ›
          Gene Symbol:
          RIMS1
          Gene Name:
          regulating synaptic membrane exocytosis 1
          Gene Aliases:
          CORD7, RAB3IP2, RIM, RIM1
          Chromosome Location:
          Chr.6: 71886703 - 72403143 on Build GRCh38
          UniGene ID:
          Hs.485729
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 136691

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RIMS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001168407.1 NP_001161879.1 3 537
          NM_001168408.1 NP_001161880.1 3 537
          NM_001168409.1 NP_001161881.1 3 322
          NM_001168410.1 NP_001161882.1 3 609
          NM_014989.5 NP_055804.2 8 2124
          XM_005248688.3 XP_005248745.1 8 2060
          XM_011535604.2 XP_011533906.2 8 2060
          XM_011535611.2 XP_011533913.2 8 2060
          XM_011535615.2 XP_011533917.2 8 2060
          XM_017010516.1 XP_016866005.1 8 2060
          XM_017010517.1 XP_016866006.1 8 2060
          XM_017010518.1 XP_016866007.1 8 2060
          XM_017010519.1 XP_016866008.1 8 2060
          XM_017010520.1 XP_016866009.1 8 2060
          XM_017010521.1 XP_016866010.1 8 2060
          XM_017010522.1 XP_016866011.1 8 2060
          XM_017010523.1 XP_016866012.1 8 2060
          XM_017010524.1 XP_016866013.1 8 2060
          XM_017010525.1 XP_016866014.1 8 2060
          XM_017010526.1 XP_016866015.1 8 2060
          XM_017010527.1 XP_016866016.1 8 2060
          XM_017010528.1 XP_016866017.1 8 2060
          XM_017010529.1 XP_016866018.1 8 2060
          XM_017010530.1 XP_016866019.1 8 2060
          XM_017010531.1 XP_016866020.1 8 2060
          XM_017010532.1 XP_016866021.1 8 2060
          XM_017010533.1 XP_016866022.1 8 2060
          XM_017010534.1 XP_016866023.1 3 633
          XM_017010535.1 XP_016866024.1 3 633
          XM_017010536.1 XP_016866025.1 3 630
          XM_017010537.1 XP_016866026.1 3 630
          XM_017010538.1 XP_016866027.1 3 738
          XM_017010539.1 XP_016866028.1 3 625
          XM_017010540.1 XP_016866029.1 3 624
          XM_017010541.1 XP_016866030.1 3 624
          XM_017010542.1 XP_016866031.1 3 624
          XM_017010543.1 XP_016866032.1 3 543
          XM_017010544.1 XP_016866033.1 3 626
          XM_017010545.1 XP_016866034.1 3 623
          XM_017010546.1 XP_016866035.1 3 736
          XM_017010547.1 XP_016866036.1 3 736
          XM_017010548.1 XP_016866037.1 3 621
          XM_017010549.1 XP_016866038.1 3 621
          XM_017010550.1 XP_016866039.1 3 1004
          XM_017010551.1 XP_016866040.1 3 1004
          XM_017010552.1 XP_016866041.1 3 861
          XM_017010553.1 XP_016866042.1 3 861
          XM_017010554.1 XP_016866043.1 3 857
          XM_017010555.1 XP_016866044.1 3 857
          GenBank mRNA AB002338.1 - 9 1847
          AB045726.1 - 9 2090
          AB051866.1 - 9 2090
          AK293828.1 - 3 212
          AK294868.1 - 3 537
          AK295001.1 - 3 455
          AK295177.1 - 3 523
          AK296303.1 - 3 378
          AK308658.1 - 3 609
          AK309185.1 - 3 322
          AK311105.1 - 3 484
          AK316108.1 - 3 335
          AY190519.1 - 8 1803
          BC151853.1 - 9 1871 8860502
          BC152435.1 - 9 1847 8862556

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          protein complex assembly
          intracellular protein transport
          neurotransmitter secretion
          visual perception
          positive regulation of gene expression
          glutamate secretion
          synaptic vesicle exocytosis
          calcium ion regulated exocytosis
          regulated exocytosis
          secretion
          regulation of neurotransmitter secretion
          regulation of catalytic activity
          response to stimulus
          long-term synaptic potentiation
          membrane fusion
          positive regulation of inhibitory postsynaptic potential
          positive regulation of dendrite extension
          positive regulation of excitatory postsynaptic potential
          protein binding
          SH3 domain binding
          Rab GTPase binding
          GTPase regulator activity
          poly(A) RNA binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0