Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SYNE2 SIRNA Assays ›
Gene Symbol:
SYNE2
Gene Name:
spectrin repeat containing nuclear envelope protein 2
Gene Aliases:
EDMD5, NUA, NUANCE, Nesp2, Nesprin-2, SYNE-2, TROPH
Chromosome Location:
Chr.14: 63761904 - 64226449 on Build GRCh38
UniGene ID:
Hs.745014
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
136818
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
286.00
Your Price
Online offer:
277.65
277.65
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SYNE2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_015180.4
NP_055995.4
109
19903
NM_182910.2
NP_878914.1
3
634
NM_182913.2
NP_878917.1
5
739
NM_182914.2
NP_878918.2
110
19972
XM_005267454.1
XP_005267511.1
110
19972
XM_005267456.1
XP_005267513.1
110
19972
XM_005267457.1
XP_005267514.1
110
19972
XM_005267458.1
XP_005267515.1
109
19903
XM_005267459.1
XP_005267516.1
109
19903
XM_011536574.1
XP_011534876.1
110
20014
XM_011536575.2
XP_011534877.1
110
19991
XM_011536576.2
XP_011534878.1
111
20171
XM_011536577.2
XP_011534879.1
110
19938
XM_011536578.1
XP_011534880.1
110
20014
XM_011536579.1
XP_011534881.1
110
20014
XM_011536580.1
XP_011534882.1
110
20014
XM_011536581.1
XP_011534883.1
109
19945
XM_011536582.1
XP_011534884.1
109
19897
XM_017021101.1
XP_016876590.1
110
20016
XM_017021102.1
XP_016876591.1
109
20059
XM_017021103.1
XP_016876592.1
12
2051
XM_017021104.1
XP_016876593.1
12
2051
GenBank mRNA
AB023228.3
-
61
9644
AF357236.1
-
109
19442
AF435011.1
-
109
19885
AF495911.1
-
109
19870
AK074055.1
-
21
3370
AK090430.1
-
27
4279
AK297874.1
-
5
700
AK298465.1
-
4
414
AL080133.1
-
15
2416
AL117404.1
-
3
1156
AY061757.1
-
10
1507
AY061758.1
-
5
736
AY061759.1
-
57
9206
AY184204.1
-
3
1156
AY184205.1
-
8
1388
BX537642.1
-
30
4717
BX538095.1
-
18
2837
BX640974.1
-
11
1718
CR749324.1
-
30
4835
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
nuclear envelope organization
nuclear migration
establishment or maintenance of cell polarity
fibroblast migration
positive regulation of cell migration
nuclear migration along microfilament
protein localization to nucleus
centrosome localization
cytoskeletal anchoring at nuclear membrane
actin binding
protein binding
actin filament binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0