Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PUM2 SIRNA Assays ›
          Gene Symbol:
          PUM2
          Gene Name:
          pumilio RNA binding family member 2
          Gene Aliases:
          PUMH2, PUML2
          Chromosome Location:
          Chr.2: 20248692 - 20352423 on Build GRCh38
          UniGene ID:
          Hs.467824
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 136882

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PUM2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001282752.1 NP_001269681.1 3 449
          NM_001282790.1 NP_001269719.1 3 312
          NM_001282791.1 NP_001269720.1 4 437
          NM_015317.2 NP_056132.1 4 558
          XM_005262607.1 XP_005262664.1 4 532
          XM_005262609.1 XP_005262666.1 4 532
          XM_005262610.1 XP_005262667.1 4 532
          XM_006711973.1 XP_006712036.1 4 329
          XM_006711974.2 XP_006712037.1 4 369
          XM_006711975.2 XP_006712038.1 5 493
          XM_011532719.1 XP_011531021.1 6 728
          XM_011532720.2 XP_011531022.1 6 737
          XM_011532721.2 XP_011531023.1 5 529
          XM_011532723.1 XP_011531025.1 4 329
          XM_011532724.2 XP_011531026.1 4 532
          XM_011532725.2 XP_011531027.1 4 532
          XM_017003701.1 XP_016859190.1 4 329
          XM_017003702.1 XP_016859191.1 4 369
          XM_017003703.1 XP_016859192.1 5 557
          XM_017003704.1 XP_016859193.1 7 875
          XM_017003705.1 XP_016859194.1 7 862
          XM_017003706.1 XP_016859195.1 6 654
          XM_017003707.1 XP_016859196.1 4 329
          XM_017003708.1 XP_016859197.1 4 562
          XM_017003709.1 XP_016859198.1 5 528
          XM_017003710.1 XP_016859199.1 4 378
          XM_017003711.1 XP_016859200.1 4 329
          XM_017003712.1 XP_016859201.1 4 397
          XM_017003713.1 XP_016859202.1 5 556
          XM_017003714.1 XP_016859203.1 1 108
          XM_017003715.1 XP_016859204.1 4 329
          GenBank mRNA AF315591.1 - 3 309
          AK093847.1 - 4 410
          AK304198.1 - 3 219
          BC112046.1 - 3 312 8327514
          BC112048.1 - 3 312 8327516
          BC143550.1 - 3 312 9052061
          BC143552.1 - 4 437
          BG717417.1 - 4 565
          D87078.3 - 3 286

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001106715, XM_006239884, XM_008764582, XM_006239883, XM_008764580, and others.
          This siRNA also targets the following transcripts in Mouse: NM_030723, NM_001160220, NM_001160221, NM_001160222, NM_001160219, and others.

          Panther Classification:

          Molecular Function -

          RNA metabolism protein nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          posttranscriptional regulation of gene expression
          stress granule assembly
          regulation of mRNA stability
          regulation of chromosome segregation
          regulation of gene silencing by miRNA
          positive regulation of RIG-I signaling pathway
          positive regulation of gene silencing by miRNA
          RNA binding
          mRNA 3'-UTR binding
          protein binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0