Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CELF1 SIRNA Assays ›
Gene Symbol:
CELF1
Gene Name:
CUGBP, Elav-like family member 1
Gene Aliases:
BRUNOL2, CUG-BP, CUGBP, CUGBP1, EDEN-BP, NAB50, NAPOR, hNab50
Chromosome Location:
Chr.11: 47465937 - 47565569 on Build GRCh38
UniGene ID:
Hs.595333
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
137105
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CELF1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001025596.2
NP_001020767.1
6
556
NM_001172639.1
NP_001166110.1
9
1518
NM_001172640.1
NP_001166111.1
6
553
NM_006560.3
NP_006551.1
7
805
NM_198700.2
NP_941989.1
7
747
XM_011519848.2
XP_011518150.1
7
628
XM_011519850.2
XP_011518152.2
7
867
XM_011519852.2
XP_011518154.1
9
976
XM_011519854.1
XP_011518156.1
11
1084
XM_011519855.1
XP_011518157.1
7
805
XM_011519856.1
XP_011518158.1
8
886
XM_011519857.1
XP_011518159.1
7
757
XM_011519858.1
XP_011518160.1
7
852
XM_011519859.1
XP_011518161.1
11
1084
XM_017017101.1
XP_016872590.1
10
968
XM_017017102.1
XP_016872591.1
9
887
XM_017017103.1
XP_016872592.1
8
780
XM_017017104.1
XP_016872593.1
8
780
XM_017017105.1
XP_016872594.1
8
780
XM_017017106.1
XP_016872595.1
10
968
XM_017017107.1
XP_016872596.1
8
780
XM_017017108.1
XP_016872597.1
8
780
XM_017017109.1
XP_016872598.1
7
867
XM_017017110.1
XP_016872599.1
8
780
XM_017017111.1
XP_016872600.1
8
780
XM_017017112.1
XP_016872601.1
8
780
XM_017017113.1
XP_016872602.1
8
780
XM_017017114.1
XP_016872603.1
8
780
XM_017017115.1
XP_016872604.1
9
808
XM_017017116.1
XP_016872605.1
8
843
XM_017017117.1
XP_016872606.1
7
604
XM_017017118.1
XP_016872607.1
7
726
XM_017017119.1
XP_016872608.1
9
945
XM_017017120.1
XP_016872609.1
7
757
XM_017017121.1
XP_016872610.1
7
852
XM_017017122.1
XP_016872611.1
7
757
XM_017017123.1
XP_016872612.1
7
805
XM_017017124.1
XP_016872613.1
9
899
XM_017017125.1
XP_016872614.1
7
628
XM_017017126.1
XP_016872615.1
7
757
XM_017017127.1
XP_016872616.1
7
781
XM_017017128.1
XP_016872617.1
7
805
XM_017017129.1
XP_016872618.1
8
803
XM_017017130.1
XP_016872619.1
7
757
XM_017017131.1
XP_016872620.1
7
781
XM_017017132.1
XP_016872621.1
7
805
XM_017017133.1
XP_016872622.1
7
596
XM_017017134.1
XP_016872623.1
8
735
XM_017017135.1
XP_016872624.1
7
757
GenBank mRNA
AB210019.1
-
9
1518
AF248648.1
-
6
564
AF267533.1
-
6
557
AF267534.1
-
6
557
AJ007988.1
-
6
546
AK304430.1
-
6
496
AK307679.1
-
1
18
BC031079.1
-
7
747
5297141
U63289.1
-
7
683
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
mRNA splice site selection
mRNA processing
germ cell development
spermatid development
embryo development
RNA interference
negative regulation of translation
positive regulation of multicellular organism growth
regulation of RNA splicing
nucleotide binding
translation repressor activity, nucleic acid binding
RNA binding
mRNA binding
protein binding
translation initiation factor binding
BRE binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0