Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PRPF4B SIRNA Assays ›
          Gene Symbol:
          PRPF4B
          Gene Name:
          pre-mRNA processing factor 4B
          Gene Aliases:
          PR4H, PRP4, PRP4H, PRP4K, dJ1013A10.1
          Chromosome Location:
          Chr.6: 4019402 - 4064983 on Build GRCh38
          UniGene ID:
          Hs.159014
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 137361

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PRPF4B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_003913.4 NP_003904.3 2 450
          XM_011514970.2 XP_011513272.1 2 2383
          XM_017011410.1 XP_016866899.1 2 2383
          XM_017011411.1 XP_016866900.1 2 2383
          XM_017011412.1 XP_016866901.1 2 2383
          XM_017011413.1 XP_016866902.1 2 2383
          XM_017011414.1 XP_016866903.1 2 2383
          XM_017011415.1 XP_016866904.1 2 2383
          XM_017011416.1 XP_016866905.1 2 2383
          XM_017011417.1 XP_016866906.1 2 2383
          XM_017011418.1 XP_016866907.1 2 2383
          XM_017011419.1 XP_016866908.1 2 2383
          XM_017011420.1 XP_016866909.1 2 2383
          XM_017011421.1 XP_016866910.1 2 2383
          XM_017011422.1 XP_016866911.1 2 2383
          XM_017011423.1 XP_016866912.1 2 2383
          XM_017011424.1 XP_016866913.1 2 2383
          XM_017011425.1 XP_016866914.1 2 2383
          XM_017011426.1 XP_016866915.1 2 2383
          XM_017011427.1 XP_016866916.1 2 2383
          XM_017011428.1 XP_016866917.1 2 2383
          XM_017011429.1 XP_016866918.1 2 2383
          XM_017011430.1 XP_016866919.1 2 2383
          XM_017011431.1 XP_016866920.1 2 2383
          XM_017011432.1 XP_016866921.1 2 2383
          XM_017011433.1 XP_016866922.1 2 2383
          XM_017011434.1 XP_016866923.1 2 2383
          XM_017011435.1 XP_016866924.1 2 2383
          GenBank mRNA AB011108.1 - 2 423
          AF283465.1 - 2 401
          AK291244.1 - 2 428
          AK303047.1 - 2 475
          AY029347.1 - 2 359
          BC034969.1 - 2 463 4821304
          CN314761.1 - 2 411

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA splicing, via spliceosome
          protein phosphorylation
          RNA splicing
          protein kinase activity
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0