Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SGCE SIRNA Assays ›
Gene Symbol:
SGCE
Gene Name:
sarcoglycan epsilon
Gene Aliases:
DYT11, ESG
Chromosome Location:
Chr.7: 94585224 - 94656209 on Build GRCh38
UniGene ID:
Hs.371199
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
137372
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SGCE
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001099400.1
NP_001092870.1
3, 4
494
NM_001099401.1
NP_001092871.1
3, 4
494
NM_001301139.1
NP_001288068.1
2, 3
371
NM_003919.2
NP_003910.1
3, 4
494
XM_005250675.4
XP_005250732.1
4, 5
596
XM_005250677.4
XP_005250734.1
4, 5
577
XM_011516663.2
XP_011514965.1
4, 5
577
XM_011516664.2
XP_011514966.1
4, 5
577
XM_011516665.2
XP_011514967.1
3, 4
472
XM_011516666.2
XP_011514968.1
3, 4
472
XM_011516667.2
XP_011514969.1
5, 6
609
XM_011516669.2
XP_011514971.1
4, 5
515
XM_017012763.1
XP_016868252.1
4, 5
445
XM_017012764.1
XP_016868253.1
3, 4
472
XM_017012765.1
XP_016868254.1
5, 6
608
XM_017012766.1
XP_016868255.1
4, 5
445
XM_017012767.1
XP_016868256.1
4, 5
528
XM_017012768.1
XP_016868257.1
4, 5
515
XM_017012769.1
XP_016868258.1
4, 5
515
XM_017012770.1
XP_016868259.1
4, 5
574
GenBank mRNA
AB117974.1
-
3, 4
346
AF036364.1
-
3, 4
422
AF516515.1
-
3, 4
383
AJ000534.1
-
3, 4
451
AK295021.1
-
4, 5
559
AK298220.1
-
2, 3
328
AK313438.1
-
3, 4
433
AY359042.1
-
3, 4
400
BC021709.2
-
3, 4
434
4822278
BI552969.1
-
3, 4
499
KU178449.1
-
3, 4
383
KU178450.1
-
3, 4
383
Back To Top
More Information
Panther Classification:
Molecular Function -
cadherin
Gene Ontology Categories:
Function(s)
muscle system process
cell-matrix adhesion
muscle organ development
membrane organization
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0