Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other WASF1 SIRNA Assays ›
          Gene Symbol:
          WASF1
          Gene Name:
          WAS protein family member 1
          Gene Aliases:
          SCAR1, WAVE, WAVE1
          Chromosome Location:
          Chr.6: 110099817 - 110189874 on Build GRCh38
          UniGene ID:
          Hs.75850
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 137386

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: WASF1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001024934.1 NP_001020105.1 4 961
          NM_001024935.1 NP_001020106.1 4 914
          NM_001024936.1 NP_001020107.1 3 816
          NM_003931.2 NP_003922.1 5 1059
          XM_005267203.3 XP_005267260.1 6 1057
          XM_005267205.2 XP_005267262.1 4 523
          XM_005267206.2 XP_005267263.1 4 571
          XM_005267207.2 XP_005267264.1 3 386
          XM_011536233.1 XP_011534535.1 4 671
          XM_011536234.1 XP_011534536.1 5 959
          XM_011536235.1 XP_011534537.1 5 1017
          XM_017011437.1 XP_016866926.1 6 1079
          XM_017011438.1 XP_016866927.1 6 1021
          XM_017011439.1 XP_016866928.1 4 612
          XM_017011440.1 XP_016866929.1 3 489
          XM_017011441.1 XP_016866930.1 4 523
          XM_017011442.1 XP_016866931.1 4 817
          XM_017011443.1 XP_016866932.1 3 419
          XM_017011444.1 XP_016866933.1 4 571
          GenBank mRNA AF134303.1 - 2 222
          AK097838.1 - 5 610
          BC044591.1 - 4 532 5265612
          D87459.1 - 4 464

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-motor actin binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          protein complex assembly
          movement of cell or subcellular component
          Rac protein signal transduction
          actin filament polymerization
          lamellipodium morphogenesis
          positive regulation of Arp2/3 complex-mediated actin nucleation
          actin binding
          protein binding
          protein complex binding
          Rac GTPase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0