Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MTRF1 SIRNA Assays ›
          Gene Symbol:
          MTRF1
          Gene Name:
          mitochondrial translational release factor 1
          Gene Aliases:
          MRF1, MTTRF1, RF1
          Chromosome Location:
          Chr.13: 41216369 - 41311794 on Build GRCh38
          UniGene ID:
          Hs.382176
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 137556

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MTRF1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_004294.2 NP_004285.2 9, 10 1333
          XM_005266599.2 XP_005266656.1 11, 12 1582
          XM_006719897.2 XP_006719960.1 11, 12 1690
          XM_006719898.2 XP_006719961.1 11, 12 1674
          XM_006719900.2 XP_006719963.1 10, 11 1403
          XM_011535315.2 XP_011533617.1 11, 12 1799
          XM_011535317.2 XP_011533619.1 10, 11 1682
          XM_011535318.2 XP_011533620.1 11, 12 1842
          XM_011535320.2 XP_011533622.1 11, 12 1778
          XM_011535321.2 XP_011533623.1 14, 15 1914
          XM_011535323.2 XP_011533625.1 11, 12 1644
          XM_011535325.2 XP_011533627.1 10, 11 1597
          XM_017020856.1 XP_016876345.1 10, 11 1643
          XM_017020857.1 XP_016876346.1 13, 14 1777
          XM_017020858.1 XP_016876347.1 12, 13 1772
          XM_017020860.1 XP_016876349.1 12, 13 1660
          XM_017020862.1 XP_016876351.1 11, 12 2855
          XM_017020863.1 XP_016876352.1 11, 12 2703
          XM_017020864.1 XP_016876353.1 11, 12 2595
          XM_017020865.1 XP_016876354.1 10, 11 2547
          XM_017020866.1 XP_016876355.1 9, 10 2412
          XM_017020867.1 XP_016876356.1 11, 12 1560
          XM_017020868.1 XP_016876357.1 9, 10 1528
          XM_017020869.1 XP_016876358.1 10, 11 1426
          XM_017020873.1 XP_016876362.1 14, 15 1984
          XM_017020874.1 XP_016876363.1 11, 12 1536
          XM_017020875.1 XP_016876364.1 11, 12 1530
          XM_017020876.1 XP_016876365.1 10, 11 2870
          XM_017020877.1 XP_016876366.1 12, 13 1705
          GenBank mRNA AF072934.1 - 9, 10 1304
          AK055086.1 - 11, 12 1704
          BC042196.1 - 9, 10 1333 5198308

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          translation release factor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of translational termination
          mitochondrial translational termination
          translation release factor activity
          translation release factor activity, codon specific
          ribosome binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0