Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NPIPA3 SIRNA Assays ›
          Gene Symbol:
          NPIPA3 NPIPA1 NPIPA2 NPIPA5 NPIPA7 NPIPA8 PKD1P1 PKD1P6-NPIPP1
          Gene Name:
          nuclear pore complex interacting protein family member A3
          nuclear pore complex interacting protein family member A1
          nuclear pore complex interacting protein family member A2
          nuclear pore complex interacting protein family member A5
          nuclear pore complex interacting protein family member A7
          nuclear pore complex interacting protein family member A8
          polycystin 1, transient receptor potential channel interacting pseudogene 1
          PKD1P6-NPIPP1 readthrough
          Gene Aliases:
          -
          NPIP, NPIPA, morpheus
          NPIP
          NPIP
          -
          LCR16a9
          HG1, NPIP
          -
          Chromosome Location:
          Chr.16: 14706473 - 14726338 on Build GRCh38
          Chromosome Location:
          Chr.16: 14935258 - 14952074 on Build GRCh38
          Chromosome Location:
          Chr.16: 14745595 - 14765448 on Build GRCh38
          Chromosome Location:
          Chr.16: 15363624 - 15381478 on Build GRCh38
          Chromosome Location:
          Chr.16: 16376940 - 16393972 on Build GRCh38
          Chromosome Location:
          Chr.16: 18317919 - 18334967 on Build GRCh38
          Chromosome Location:
          Chr.16: 16317609 - 16350608 on Build GRCh38
          Chromosome Location:
          Chr.16: 15104723 - 15131601 on Build GRCh38
          UniGene ID:
          -
          Hs.626677
          -
          Hs.676266
          Hs.634612
          Hs.636569
          Hs.636569
          Hs.634612
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID 138198

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NPIPA3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001277323.1 NP_001264252.1 5 460
          XM_005255484.3 XP_005255541.1 6 548
          XM_005255485.4 XP_005255542.1 5 864
          XM_011522576.2 XP_011520878.2 7 2913
          XM_011522577.2 XP_011520879.2 7 2834
          XM_011522578.2 XP_011520880.2 5 556
          XM_011522580.2 XP_011520882.2 5 487
          XM_011522581.1 XP_011520883.1 7 2923
          XM_011522583.2 XP_011520885.1 7 2923
          XM_011522584.1 XP_011520886.1 7 2923
          XM_017023553.1 XP_016879042.1 7 2923
          XM_017023554.1 XP_016879043.1 6 2836
          Gene Symbol: NPIPA1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_006985.2 NP_008916.2 5 460
          XM_011522748.2 XP_011521050.1 5 557
          XM_011522749.2 XP_011521051.1 6 669
          XM_011522751.1 XP_011521053.1 5 556
          XM_017023891.1 XP_016879380.1 6 669
          GenBank mRNA AF132984.1 - 5 460
          AK160377.1 - 24 4647
          KF175165.1 - 5 496
          KF175167.1 - 5 496
          KF175169.1 - 5 460
          KF175208.1 - 5 607
          KF175209.1 - 6 678
          KF175225.1 - 5 460
          Gene Symbol: NPIPA2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001277324.1 NP_001264253.1 5 460
          XM_005255489.3 XP_005255546.1 7 2879
          XM_005255492.4 XP_005255549.1 5 864
          XM_006720917.3 XP_006720980.2 7 2923
          XM_011522587.1 XP_011520889.1 7 2923
          XM_011522593.1 XP_011520895.1 7 2923
          XM_011522594.2 XP_011520896.1 7 2923
          XM_011522595.1 XP_011520897.1 7 2923
          GenBank mRNA KF175213.1 - 4 153
          Gene Symbol: NPIPA5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001277325.1 NP_001264254.1 5 460
          XM_011522329.2 XP_011520631.1 7 871
          XM_011522336.2 XP_011520638.1 7 864
          GenBank mRNA CB123100.1 - 2 138
          EL734608.1 - 5 460
          KF175168.1 - 5 457
          KF175170.1 - 5 460
          KF175171.1 - 5 454
          KF175172.1 - 5 460
          KF175173.1 - 3 235
          Gene Symbol: NPIPA7
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001282507.1 NP_001269436.1 5 460
          XM_005255735.4 XP_005255792.1 5 582
          XM_011522339.2 XP_011520641.1 5 508
          XM_017022833.1 XP_016878322.1 6 533
          XM_017022834.1 XP_016878323.1 6 585
          XM_017022835.1 XP_016878324.1 6 646
          XM_017022836.1 XP_016878325.1 5 453
          XM_017022837.1 XP_016878326.1 6 533
          XM_017022838.1 XP_016878327.1 5 928
          XM_017022840.1 XP_016878329.1 5 473
          GenBank mRNA BI046418.1 - 2 111
          Gene Symbol: NPIPA8
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001282511.1 NP_001269440.1 5 460
          XM_005255738.4 XP_005255795.1 5 582
          XM_006720830.3 XP_006720893.1 5 1480
          XM_011522342.2 XP_011520644.1 5 509
          XM_017022841.1 XP_016878330.1 6 533
          XM_017022842.1 XP_016878331.1 6 585
          XM_017022843.1 XP_016878332.1 6 646
          XM_017022844.1 XP_016878333.1 5 453
          XM_017022845.1 XP_016878334.1 6 533
          XM_017022847.1 XP_016878336.1 5 473
          GenBank mRNA BQ101079.1 - 5 506
          BY996642.1 - 1 41
          Gene Symbol: PKD1P1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK298381.1 - 3 218
          BC039707.1 - 6 779 4479933
          BC046145.1 - 7 871
          KF175174.1 - 5 496
          KF175211.1 - 7 816
          KF175212.1 - 8 629
          KF175214.1 - 6 678
          KF175219.1 - 5 646
          Gene Symbol: PKD1P6-NPIPP1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_123721.1 24 4553
          NR_123722.1 14 2414
          GenBank mRNA AK131084.1 - 19 4917
          AK131085.1 - 18 4823
          AK302708.1 - 14 2414

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          protein transport
          mRNA transport
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0