Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPP6R2 SIRNA Assays ›
Gene Symbol:
PPP6R2
Gene Name:
protein phosphatase 6 regulatory subunit 2
Gene Aliases:
KIAA0685, PP6R2, SAP190, SAPS2
Chromosome Location:
Chr.22: 50343317 - 50445090 on Build GRCh38
UniGene ID:
Hs.449098
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
138324
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPP6R2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242898.1
NP_001229827.1
9
1265
NM_001242899.1
NP_001229828.1
9
1268
NM_001242900.1
NP_001229829.1
9
1265
NM_014678.4
NP_055493.2
9
1265
XM_005261960.3
XP_005262017.1
3
219
XM_006724431.2
XP_006724494.1
10
1376
XM_006724434.1
XP_006724497.1
11
1525
XM_006724435.2
XP_006724498.1
9
1250
XM_011530720.2
XP_011529022.1
8
953
XM_011530721.2
XP_011529023.1
8
950
XM_011530722.2
XP_011529024.1
8
950
XM_011530723.2
XP_011529025.1
8
953
XM_011530724.2
XP_011529026.1
8
953
XM_011530726.2
XP_011529028.1
8
953
XM_011530727.2
XP_011529029.1
8
953
XM_011530728.2
XP_011529030.1
8
953
XM_011530729.2
XP_011529031.1
8
953
XM_011530730.2
XP_011529032.1
8
950
XM_011530731.1
XP_011529033.1
8
950
XM_011530732.2
XP_011529034.1
11
1429
XM_011530734.2
XP_011529036.1
10
1303
XM_011530735.2
XP_011529037.1
9
1253
XM_011530736.2
XP_011529038.1
10
1379
XM_011530737.2
XP_011529039.1
11
1528
XM_011530738.2
XP_011529040.1
10
1331
XM_011530739.2
XP_011529041.1
8
993
XM_011530740.2
XP_011529042.1
10
1401
XM_017029116.1
XP_016884605.1
8
950
XM_017029117.1
XP_016884606.1
12
1578
XM_017029118.1
XP_016884607.1
12
1530
XM_017029119.1
XP_016884608.1
11
1452
XM_017029120.1
XP_016884609.1
11
1426
XM_017029121.1
XP_016884610.1
9
1250
XM_017029122.1
XP_016884611.1
10
1328
XM_017029123.1
XP_016884612.1
9
1250
XM_017029124.1
XP_016884613.1
10
1376
XM_017029125.1
XP_016884614.1
9
1250
XM_017029126.1
XP_016884615.1
9
1253
XM_017029127.1
XP_016884616.1
10
1328
XM_017029128.1
XP_016884617.1
10
1376
XM_017029129.1
XP_016884618.1
9
1250
XM_017029130.1
XP_016884619.1
9
1250
XM_017029131.1
XP_016884620.1
10
1376
XM_017029132.1
XP_016884621.1
10
1328
XM_017029133.1
XP_016884622.1
9
1207
XM_017029134.1
XP_016884623.1
9
1204
XM_017029135.1
XP_016884624.1
10
1254
XM_017029136.1
XP_016884625.1
9
1204
XM_017029137.1
XP_016884626.1
9
1204
XM_017029138.1
XP_016884627.1
9
1204
GenBank mRNA
AB014585.1
-
9
1237
AK002063.1
-
2
61
AK302472.1
-
9
1176
BC000976.4
-
10
1254
2900437
BC006568.2
-
9
1251
3532211
BC032664.1
-
9
1265
5553142
BC041698.1
-
9
1235
5732101
BC052995.2
-
9
1171
6162781
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001106789, XM_008765688, FQ230825, BC166525, FQ211682.
Panther Classification:
Molecular Function -
phosphatase modulator
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0