Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TFAP2B SIRNA Assays ›
          Gene Symbol:
          TFAP2B
          Gene Name:
          transcription factor AP-2 beta
          Gene Aliases:
          AP-2B, AP2-B
          Chromosome Location:
          Chr.6: 50817692 - 50847619 on Build GRCh38
          UniGene ID:
          Hs.33102
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 138844

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TFAP2B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_003221.3 NP_003212.2 5 1081
          XM_011514837.2 XP_011513139.1 6 1386
          XM_017011233.1 XP_016866722.1 8 1762
          XM_017011234.1 XP_016866723.1 8 1215
          XM_017011235.1 XP_016866724.1 4 894
          GenBank mRNA AK291172.1 - 5 921
          AK313749.1 - 5 919
          BC037225.1 - 5 937 4745989
          X95694.1 - 5 914

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001106896, XM_008766927, XM_006244649.
          This siRNA also targets the following transcripts in Mouse: NM_001286340, NM_009334, NM_001025305, X78197, AK032045, and others.

          Panther Classification:

          Molecular Function -

          general transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          kidney development
          renal water homeostasis
          glucose metabolic process
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          apoptotic process
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          response to lithium ion
          retina layer formation
          magnesium ion homeostasis
          regulation of BMP signaling pathway
          forelimb morphogenesis
          hindlimb morphogenesis
          positive regulation of urine volume
          aorta morphogenesis
          response to drug
          glucose homeostasis
          negative regulation of apoptotic process
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          negative regulation of neuron apoptotic process
          positive regulation of neuron apoptotic process
          skin development
          fat cell differentiation
          regulation of cell differentiation
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          sympathetic nervous system development
          regulation of insulin secretion
          phosphate ion homeostasis
          calcium ion homeostasis
          potassium ion homeostasis
          sodium ion homeostasis
          distal tubule development
          collecting duct development
          metanephric nephron development
          ductus arteriosus closure
          cellular ammonia homeostasis
          cellular creatinine homeostasis
          cellular urea homeostasis
          RNA polymerase II core promoter sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          transcription factor activity, RNA polymerase II core promoter sequence-specific
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          RNA polymerase II transcription coactivator activity
          RNA polymerase II transcription corepressor activity
          enhancer sequence-specific DNA binding
          DNA binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          transcription coactivator activity
          transcription corepressor activity
          protein binding
          protein homodimerization activity
          cysteine-type endopeptidase inhibitor activity involved in apoptotic process
          sequence-specific DNA binding
          protein heterodimerization activity
          protein dimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0