Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TGFB3 SIRNA Assays ›
Gene Symbol:
TGFB3
Gene Name:
transforming growth factor beta 3
Gene Aliases:
ARVD, ARVD1, LDS5, RNHF, TGF-beta3
Chromosome Location:
Chr.14: 75958097 - 75982022 on Build GRCh38
UniGene ID:
Hs.592317
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
138855
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TGFB3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_003239.3
NP_003230.1
7
3232
GenBank mRNA
AK122902.1
-
4
2484
J03241.1
-
7
2363
X14149.1
-
7
2350
Back To Top
More Information
Panther Classification:
Molecular Function -
growth factor
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPK activity
response to hypoxia
in utero embryonic development
platelet degranulation
transforming growth factor beta receptor signaling pathway
SMAD protein import into nucleus
salivary gland morphogenesis
female pregnancy
aging
negative regulation of DNA replication
negative regulation of cell proliferation
positive regulation of epithelial to mesenchymal transition
positive regulation of pathway-restricted SMAD protein phosphorylation
negative regulation of macrophage cytokine production
cell growth
positive regulation of bone mineralization
negative regulation of transforming growth factor beta receptor signaling pathway
mammary gland development
response to progesterone
positive regulation of collagen biosynthetic process
response to laminar fluid shear stress
wound healing
regulation of cell proliferation
odontogenesis
uterine wall breakdown
regulation of apoptotic process
positive regulation of apoptotic process
regulation of MAPK cascade
negative regulation of neuron apoptotic process
response to estrogen
ossification involved in bone remodeling
cell-cell junction organization
positive regulation of DNA replication
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
lung alveolus development
digestive tract development
embryonic neurocranium morphogenesis
inner ear development
positive regulation of protein secretion
positive regulation of filopodium assembly
positive regulation of stress fiber assembly
positive regulation of cell division
palate development
face morphogenesis
frontal suture morphogenesis
positive regulation of SMAD protein import into nucleus
SMAD protein signal transduction
detection of hypoxia
regulation of epithelial to mesenchymal transition involved in endocardial cushion formation
positive regulation of occluding junction disassembly
type II transforming growth factor beta receptor binding
cytokine activity
protein binding
growth factor activity
type I transforming growth factor beta receptor binding
type III transforming growth factor beta receptor binding
identical protein binding
protein heterodimerization activity
transforming growth factor beta binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0