Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other THBS1 SIRNA Assays ›
          Gene Symbol:
          THBS1
          Gene Name:
          thrombospondin 1
          Gene Aliases:
          THBS, THBS-1, TSP, TSP-1, TSP1
          Chromosome Location:
          Chr.15: 39581079 - 39598918 on Build GRCh38
          UniGene ID:
          Hs.164226
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 138863

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: THBS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_003246.3 NP_003237.2 22 4901
          XM_011521971.1 XP_011520273.1 21 4723
          GenBank mRNA AU117102.1 - 1 446
          BC136469.1 - 22 4875 9020458
          BC136470.1 - 22 4875 9020459
          BG251169.1 - 1 24
          X14787.1 - 22 4836

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPK activity
          response to hypoxia
          negative regulation of endothelial cell proliferation
          negative regulation of cell-matrix adhesion
          sprouting angiogenesis
          chronic inflammatory response
          platelet degranulation
          negative regulation of antigen processing and presentation of peptide or polysaccharide antigen via MHC class II
          negative regulation of dendritic cell antigen processing and presentation
          inflammatory response
          immune response
          response to unfolded protein
          cell cycle arrest
          cell adhesion
          response to glucose
          positive regulation of endothelial cell migration
          negative regulation of endothelial cell migration
          negative regulation of plasma membrane long-chain fatty acid transport
          negative regulation of nitric oxide mediated signal transduction
          negative regulation of cGMP-mediated signaling
          negative regulation of plasminogen activation
          positive regulation of macrophage chemotaxis
          positive regulation of fibroblast migration
          cell migration
          negative regulation of angiogenesis
          peptide cross-linking
          positive regulation of blood coagulation
          extracellular matrix organization
          positive regulation of cell migration
          positive regulation of transforming growth factor beta receptor signaling pathway
          regulation of cGMP metabolic process
          response to magnesium ion
          response to progesterone
          negative regulation of interleukin-12 production
          positive regulation of transforming growth factor beta1 production
          cellular response to heat
          response to endoplasmic reticulum stress
          protein O-linked fucosylation
          negative regulation of fibroblast growth factor receptor signaling pathway
          positive regulation of phosphorylation
          response to drug
          positive regulation of tumor necrosis factor biosynthetic process
          positive regulation of macrophage activation
          negative regulation of apoptotic process
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          positive regulation of blood vessel endothelial cell migration
          negative regulation of blood vessel endothelial cell migration
          engulfment of apoptotic cell
          positive regulation of translation
          positive regulation of angiogenesis
          behavioral response to pain
          positive regulation of chemotaxis
          response to calcium ion
          negative regulation of focal adhesion assembly
          positive regulation of protein kinase B signaling
          negative regulation of fibrinolysis
          positive regulation of extrinsic apoptotic signaling pathway via death domain receptors
          positive regulation of endothelial cell apoptotic process
          positive regulation of reactive oxygen species metabolic process
          negative regulation of endothelial cell chemotaxis
          negative regulation of extrinsic apoptotic signaling pathway
          phosphatidylserine binding
          glycoprotein binding
          fibronectin binding
          integrin binding
          calcium ion binding
          protein binding
          heparin binding
          fibroblast growth factor binding
          low-density lipoprotein particle binding
          identical protein binding
          laminin binding
          proteoglycan binding
          transforming growth factor beta binding
          fibrinogen binding
          collagen V binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-4sktr:80/100.66.76.202:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0