Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SOCS2 SIRNA Assays ›
          Gene Symbol:
          SOCS2
          Gene Name:
          suppressor of cytokine signaling 2
          Gene Aliases:
          CIS2, Cish2, SOCS-2, SSI-2, SSI2, STATI2
          Chromosome Location:
          Chr.12: 93569822 - 93626236 on Build GRCh38
          UniGene ID:
          Hs.485572
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 13897

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SOCS2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001270467.1 NP_001257396.1 3 690
          NM_001270468.1 NP_001257397.1 3 659
          NM_001270469.1 NP_001257398.1 3 657
          NM_001270470.1 NP_001257399.1 3 929
          NM_001270471.1 NP_001257400.1 2 582
          NM_003877.4 NP_003868.1 3 800
          XM_011538929.1 XP_011537231.1 2 625
          XM_011538935.1 XP_011537237.1 2 625
          XM_011538936.1 XP_011537238.1 2 625
          XM_017020147.1 XP_016875636.1 3 773
          XM_017020148.1 XP_016875637.1 3 663
          XM_017020149.1 XP_016875638.1 3 657
          XM_017020150.1 XP_016875639.1 3 864
          XM_017020151.1 XP_016875640.1 3 669
          XM_017020152.1 XP_016875641.1 3 698
          XM_017020153.1 XP_016875642.1 3 440
          XM_017020154.1 XP_016875643.1 3 502
          XM_017020155.1 XP_016875644.1 2 625
          XM_017020156.1 XP_016875645.1 3 915
          GenBank mRNA AB004903.1 - 2 248
          AB006966.1 - 2 210
          AB451318.1 - 2 210
          AF020590.1 - 2 409
          AF037989.1 - 3 527
          AK290546.1 - 3 510
          AK297662.1 - 4 390
          AK313165.1 - 3 637
          AL522912.3 - 3 804
          BC010399.1 - 2 422 4277425
          BC070039.1 - 2 2315

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_058208, AF075382.
          This siRNA also targets the following transcripts in Mouse: NM_001168656, NM_001168655, NM_007706, NM_001168657, AK137403, and others.

          Panther Classification:

          Molecular Function -

          kinase modulator

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cell growth
          negative regulation of protein kinase activity
          JAK-STAT cascade
          regulation of signal transduction
          positive regulation of signal transduction
          protein ubiquitination
          cytokine-mediated signaling pathway
          response to estradiol
          cellular response to hormone stimulus
          negative regulation of apoptotic process
          negative regulation of JAK-STAT cascade
          negative regulation of insulin receptor signaling pathway
          growth hormone receptor signaling pathway
          protein kinase inhibitor activity
          SH3/SH2 adaptor activity
          growth hormone receptor binding
          insulin-like growth factor receptor binding
          protein binding
          JAK pathway signal transduction adaptor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0