Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other EZR SIRNA Assays ›
          Gene Symbol:
          EZR
          Gene Name:
          ezrin
          Gene Aliases:
          CVIL, CVL, HEL-S-105, VIL2
          Chromosome Location:
          Chr.6: 158765741 - 158819424 on Build GRCh38
          UniGene ID:
          Hs.487027
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 138990

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: EZR
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001111077.1 NP_001104547.1 9 1050
          NM_003379.4 NP_003370.2 8 1084
          XM_011536110.1 XP_011534412.1 5 461
          GenBank mRNA AF187552.1 - 3 297
          AF188896.1 - 3 283
          AF188897.1 - 3 279
          AF189213.1 - 3 279
          AF190059.1 - 3 271
          AF199015.1 - 3 281
          AK129727.1 - 2 107
          AK296738.1 - 9 879
          AK299456.1 - 7 913
          AK312597.1 - 9 994
          AL162086.1 - 9 1002
          BC013903.2 - 8 1001 2959399
          BC068458.1 - 8 1010 4819793
          FJ224318.1 - 8 986
          J05021.1 - 8 945
          X51521.1 - 8 986

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          intestinal D-glucose absorption
          sphingosine-1-phosphate signaling pathway
          cytoskeletal anchoring at plasma membrane
          leukocyte cell-cell adhesion
          axon guidance
          regulation of cell shape
          regulation of cell size
          positive regulation of gene expression
          protein kinase A signaling
          gland morphogenesis
          membrane to membrane docking
          microvillus assembly
          astral microtubule organization
          actin cytoskeleton reorganization
          receptor internalization
          regulation of microvillus length
          regulation of actin cytoskeleton organization
          cellular protein complex localization
          positive regulation of multicellular organism growth
          cortical microtubule organization
          establishment of epithelial cell apical/basal polarity
          filopodium assembly
          phosphatidylinositol-mediated signaling
          positive regulation of protein secretion
          negative regulation of T cell receptor signaling pathway
          actin filament bundle assembly
          establishment of centrosome localization
          establishment of endothelial barrier
          negative regulation of ERK1 and ERK2 cascade
          cellular response to cAMP
          protein localization to cell cortex
          establishment of protein localization to plasma membrane
          positive regulation of establishment of protein localization to plasma membrane
          negative regulation of interleukin-2 secretion
          regulation of NIK/NF-kappaB signaling
          regulation of organelle assembly
          terminal web assembly
          positive regulation of protein localization to early endosome
          positive regulation of cellular protein catabolic process
          negative regulation of p38MAPK cascade
          positive regulation of early endosome to late endosome transport
          actin binding
          protein binding
          microtubule binding
          protein domain specific binding
          protein kinase A catalytic subunit binding
          protein kinase A regulatory subunit binding
          S100 protein binding
          poly(A) RNA binding
          cell adhesion molecule binding
          actin filament binding
          ATPase binding
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-qvstf:80/100.66.73.196:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0