Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZFX SIRNA Assays ›
Gene Symbol:
ZFX
Gene Name:
zinc finger protein, X-linked
Gene Aliases:
ZNF926
Chromosome Location:
Chr.X: 24148994 - 24216255 on Build GRCh38
UniGene ID:
Hs.336681
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
139032
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZFX
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001178084.1
NP_001171555.1
3
503
NM_001178085.1
NP_001171556.1
6
847
NM_001178095.1
NP_001171566.1
3
582
NM_003410.3
NP_003401.2
5
711
XM_005274591.4
XP_005274648.1
4
877
XM_005274592.3
XP_005274649.1
3
692
XM_005274594.4
XP_005274651.1
2
1882
XM_006724513.3
XP_006724576.1
6
1106
XM_006724515.3
XP_006724578.1
6
1108
XM_011545578.1
XP_011543880.1
6
988
XM_011545579.1
XP_011543881.1
5
776
XM_011545581.2
XP_011543883.1
5
918
XM_011545583.2
XP_011543885.1
2
432
XM_017029788.1
XP_016885277.1
6
1029
XM_017029789.1
XP_016885278.1
5
1591
XM_017029790.1
XP_016885279.1
5
996
XM_017029791.1
XP_016885280.1
4
886
XM_017029792.1
XP_016885281.1
4
765
XM_017029793.1
XP_016885282.1
3
563
XM_017029794.1
XP_016885283.1
5
817
XM_017029795.1
XP_016885284.1
2
446
XM_017029796.1
XP_016885285.1
4
598
XM_017029797.1
XP_016885286.1
6
1057
XM_017029798.1
XP_016885287.1
2
677
XM_017029799.1
XP_016885288.1
3
692
XM_017029800.1
XP_016885289.1
2
679
GenBank mRNA
AB209892.1
-
3
615
AF505615.1
-
2
404
AF505616.1
-
2
404
AF505618.1
-
2
404
AF505619.1
-
2
404
BC136308.1
-
5
699
9020295
BC136309.1
-
5
699
9020296
DA995443.1
-
3
505
M30608.1
-
4
662
X59739.1
-
3
482
X59740.1
-
6
831
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
multicellular organism development
RNA polymerase II regulatory region sequence-specific DNA binding
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0