Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other STX3 SIRNA Assays ›
          Gene Symbol:
          STX3
          Gene Name:
          syntaxin 3
          Gene Aliases:
          STX3A
          Chromosome Location:
          Chr.11: 59755059 - 59805882 on Build GRCh38
          UniGene ID:
          Hs.180711
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 139359
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # AM51331
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: STX3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001178040.1 NP_001171511.1 2 641
          NM_004177.4 NP_004168.1 2 641
          XM_005274195.3 XP_005274252.1 2 323
          XM_005274198.2 XP_005274255.1 2 323
          XM_005274200.3 XP_005274257.1 2 323
          XM_011545221.2 XP_011543523.1 2 323
          XM_017018188.1 XP_016873677.1 2 85
          XM_017018189.1 XP_016873678.1 2 85
          XM_017018190.1 XP_016873679.1 2 85
          XM_017018191.1 XP_016873680.1 2 85
          XM_017018192.1 XP_016873681.1 2 85
          XM_017018193.1 XP_016873682.1 2 85
          XM_017018194.1 XP_016873683.1 2 85
          XM_017018195.1 XP_016873684.1 2 85
          XM_017018196.1 XP_016873685.1 2 326
          GenBank mRNA AJ002076.1 - 2 94
          AK125660.1 - 2 641
          AK290251.1 - 2 210
          AK293494.1 - 2 326
          AK297419.1 - 2 248
          AK312909.1 - 2 257
          BC007405.2 - 2 284 3010338
          BC007429.2 - 2 284 3010338
          BT006666.1 - 2 94
          BX538158.1 - 3 576
          U32315.1 - 2 132
          X90581.1 - 1 64

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00188210_m1

          Panther Classification:

          Molecular Function -

          SNARE protein

          Gene Ontology Categories:

          Function(s) Process(es)

          intracellular protein transport
          positive regulation of cell proliferation
          synaptic vesicle docking
          neuron projection development
          synaptic vesicle fusion to presynaptic active zone membrane
          positive regulation of cell adhesion
          vesicle docking
          positive regulation of chemotaxis
          long-term synaptic potentiation
          positive regulation of protein localization to plasma membrane
          positive regulation of protein localization to cell surface
          SNARE binding
          SNAP receptor activity
          protein binding
          arachidonic acid binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0