Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TLE3 SIRNA Assays ›
          Gene Symbol:
          TLE3
          Gene Name:
          transducin like enhancer of split 3
          Gene Aliases:
          ESG, ESG3, GRG3, HsT18976
          Chromosome Location:
          Chr.15: 70047790 - 70097935 on Build GRCh38
          UniGene ID:
          Hs.287362
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 139469

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TLE3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001105192.2 NP_001098662.1 11, 12 2036
          NM_001282979.1 NP_001269908.1 11, 12 2036
          NM_001282980.1 NP_001269909.1 11, 12 2036
          NM_001282981.1 NP_001269910.1 11, 12 969
          NM_001282982.1 NP_001269911.1 9, 10 818
          NM_005078.3 NP_005069.2 11, 12 2036
          NM_020908.2 NP_065959.1 11, 12 2036
          XM_005254622.4 XP_005254679.1 11, 12 2084
          XM_005254623.3 XP_005254680.1 11, 12 2084
          XM_005254625.3 XP_005254682.1 11, 12 2084
          XM_005254628.3 XP_005254685.1 11, 12 2084
          XM_005254633.2 XP_005254690.1 11, 12 2054
          XM_006720665.3 XP_006720728.1 11, 12 2051
          XM_011521976.2 XP_011520278.1 11, 12 2084
          XM_011521977.2 XP_011520279.1 11, 12 2081
          XM_011521978.2 XP_011520280.1 11, 12 2084
          XM_011521979.2 XP_011520281.1 10, 11 890
          XM_011521980.2 XP_011520282.1 11, 12 2051
          XM_011521981.1 XP_011520283.1 11, 12 2084
          XM_011521982.2 XP_011520284.1 9, 10 1832
          XM_011521983.2 XP_011520285.1 9, 10 1832
          XM_017022532.1 XP_016878021.1 11, 12 2084
          GenBank mRNA AB046767.1 - 11, 12 2018
          AK096779.1 - 9, 10 818
          AK298482.1 - 11, 12 939
          AK304459.1 - 2, 3 184
          AK315058.1 - 12, 13 1364
          AY583459.1 - 11, 12 917
          AY583460.1 - 11, 12 917
          BC041831.1 - 10, 11 1233 5268249
          BC043247.2 - 11, 12 2035 5296274
          M99438.1 - 11, 12 938

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transcription cofactor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          signal transduction
          organ morphogenesis
          Wnt signaling pathway
          beta-catenin-TCF complex assembly
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0