Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CDC14B SIRNA Assays ›
          Gene Symbol:
          CDC14B CDC14C
          Gene Name:
          cell division cycle 14B
          cell division cycle 14C
          Gene Aliases:
          CDC14B3, Cdc14B1, Cdc14B2, hCDC14B
          CDC14B2, CDC14Bretro
          Chromosome Location:
          Chr.9: 96492738 - 96619830 on Build GRCh38
          Chromosome Location:
          Chr.7: 48924561 - 48927453 on Build GRCh38
          UniGene ID:
          Hs.40582
          Hs.567757
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 139921

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CDC14B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001077181.1 NP_001070649.1 3 273
          NM_003671.3 NP_003662.1 3 731
          NM_033331.2 NP_201588.1 3 731
          XM_005252288.3 XP_005252345.1 3 328
          XM_011519147.2 XP_011517449.1 3 678
          XM_011519148.2 XP_011517450.1 3 672
          XM_011519149.2 XP_011517451.1 3 676
          XM_011519150.2 XP_011517452.1 3 675
          XM_011519151.2 XP_011517453.1 3 675
          XM_011519152.2 XP_011517454.1 3 678
          XM_011519153.2 XP_011517455.1 3 674
          XM_011519156.2 XP_011517458.2 3 849
          XM_011519158.1 XP_011517460.1 3 347
          XM_011519159.2 XP_011517461.2 3 145
          XM_017015240.1 XP_016870729.1 3 678
          XM_017015241.1 XP_016870730.1 3 683
          XM_017015242.1 XP_016870731.1 3 347
          XM_017015243.1 XP_016870732.1 3 347
          XM_017015244.1 XP_016870733.1 3 328
          XM_017015245.1 XP_016870734.1 3 849
          XM_017015246.1 XP_016870735.1 3 328
          XM_017015247.1 XP_016870736.1 3 328
          XM_017015248.1 XP_016870737.1 3 294
          XM_017015249.1 XP_016870738.1 3 237
          GenBank mRNA AF023158.1 - 3 279
          AF064104.1 - 3 731
          AF064105.2 - 3 731
          AK126388.1 - 3 265
          AK225361.1 - 3 264
          DA943563.1 - 3 273
          EF611343.1 - 3 279
          Gene Symbol: CDC14C
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_003595.1 1 301
          GenBank mRNA BC028690.2 - 1 289
          BC068452.1 - 1 300

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          protein phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA repair
          protein dephosphorylation
          G2 DNA damage checkpoint
          peptidyl-tyrosine dephosphorylation
          positive regulation of ubiquitin protein ligase activity
          protein serine/threonine phosphatase activity
          protein tyrosine phosphatase activity
          protein binding
          protein tyrosine/serine/threonine phosphatase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0