Search
Search

CEMIP| Gene Symbol: CEMIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293298.1 | NP_001280227.1 | 6 | 846 | ||
| NM_001293304.1 | NP_001280233.1 | 6 | 786 | |||
| NM_018689.2 | NP_061159.1 | 5 | 687 | |||
| GenBank mRNA | AB103330.1 | - | 5 | 659 | ||
| AK000465.1 | - | 5 | 659 | |||
| AY581148.1 | - | 6 | 818 | |||
| AY581149.1 | - | 6 | 758 | |||
| AY585237.1 | - | 5 | 659 | |||
| BC020256.1 | - | 5 | 635 | 4870106 | ||