Search
Search

LOC81691| Gene Symbol: LOC81691 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144924.1 | NP_001138396.1 | 4 | 592 | ||
| NM_001199053.1 | NP_001185982.1 | 4 | 704 | |||
| NM_030941.2 | NP_112203.2 | 4 | 592 | |||
| XM_005255604.1 | XP_005255661.1 | 4 | 608 | |||
| XM_011545963.1 | XP_011544265.1 | 4 | 393 | |||
| XM_011545964.1 | XP_011544266.1 | 4 | 569 | |||
| XM_011545965.1 | XP_011544267.1 | 4 | 569 | |||
| XM_011545966.1 | XP_011544268.1 | 3 | 331 | |||
| XM_017023739.1 | XP_016879228.1 | 4 | 608 | |||
| GenBank mRNA | AF332193.1 | - | 4 | 526 | ||
| AK314458.1 | - | 4 | 451 | |||
| AL136763.2 | - | 4 | 516 | |||
| BC007646.1 | - | 4 | 538 | 3537866 | ||
| BC068503.1 | - | 4 | 394 | 5298867 | ||