Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IFFO1 SIRNA Assays ›
Gene Symbol:
IFFO1
Gene Name:
intermediate filament family orphan 1
Gene Aliases:
HOM-TES-103, IFFO
Chromosome Location:
Chr.12: 6539528 - 6556102 on Build GRCh38
UniGene ID:
Hs.15243
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
141242
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: IFFO1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001039670.2
NP_001034759.1
9
1531
NM_001193457.1
NP_001180386.1
9
1555
NM_080730.4
NP_542768.2
9
1528
NR_036467.1
9
1628
XM_006718975.3
XP_006719038.1
9
1547
XM_006718976.3
XP_006719039.1
8
1541
XM_006718977.3
XP_006719040.1
8
1538
XM_006718981.3
XP_006719044.1
10
1216
XM_011520947.2
XP_011519249.1
8
1814
XM_011520948.2
XP_011519250.1
8
1811
XM_011520949.2
XP_011519251.1
7
1805
XM_011520952.2
XP_011519254.1
9
1480
XM_017019170.1
XP_016874659.1
8
1617
XM_017019171.1
XP_016874660.1
9
1544
XM_017019172.1
XP_016874661.1
9
1483
XM_017019173.1
XP_016874662.1
8
1114
GenBank mRNA
AF124432.1
-
9
1189
AK000710.1
-
4
761
AK095106.1
-
5
1106
AK298905.1
-
9
764
AL080214.1
-
2
104
BC001790.1
-
9
1608
BC002857.2
-
8
669
BC004384.1
-
8
669
BC010431.1
-
9
1608
BC023664.2
-
9
1608
BC063004.1
-
9
806
BC071170.1
-
4
680
BC103817.1
-
4
685
BC103818.1
-
4
685
BC110387.1
-
9
1521
5183989
BC113848.1
-
4
685
BC114454.1
-
4
682
BX648701.1
-
10
1117
BX648743.1
-
10
1117
Back To Top
More Information
Panther Classification:
Molecular Function -
dehydrogenase
oxidoreductase
metabolite interconversion enzyme
Gene Ontology Categories:
Process(es)
structural molecule activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-b27fw:80/100.66.78.82:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0