Search
Search

PTAFR| Gene Symbol: PTAFR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000952.4 | NP_000943.1 | 2 | 1364 | ||
| NM_001164721.1 | NP_001158193.1 | 3 | 1308 | |||
| NM_001164722.2 | NP_001158194.1 | 2 | 1226 | |||
| NM_001164723.2 | NP_001158195.1 | 3 | 1435 | |||
| GenBank mRNA | AK290240.1 | - | 2 | 1110 | ||
| AK292053.1 | - | 3 | 1308 | |||
| AK313174.1 | - | 2 | 1095 | |||
| AY275466.1 | - | 1 | 973 | |||
| BC013816.2 | - | 1 | 1013 | 4343214 | ||
| BC063000.1 | - | 2 | 1100 | 5176867 | ||
| BT009801.1 | - | 1 | 973 | |||
| CD013879.1 | - | 3 | 1164 | |||
| D10202.1 | - | 2 | 1085 | |||
| L07334.1 | - | 1 | 973 | |||
| M76674.1 | - | 1 | 998 | |||
| M80436.1 | - | 2 | 1239 | |||
| S52624.1 | - | 2 | 1164 | |||
Set Membership: |
|