Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RARA SIRNA Assays ›
          Gene Symbol:
          RARA
          Gene Name:
          retinoic acid receptor alpha
          Gene Aliases:
          NR1B1, RAR
          Chromosome Location:
          Chr.17: 40309171 - 40357643 on Build GRCh38
          UniGene ID:
          Hs.654583
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 142156

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RARA
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000964.3 NP_000955.1 4, 5 951
          NM_001024809.3 NP_001019980.1 3, 4 1144
          NM_001145301.2 NP_001138773.1 4, 5 1063
          XM_005257552.4 XP_005257609.1 3, 4 521
          XM_005257553.1 XP_005257610.1 4, 5 855
          XM_005257554.1 XP_005257611.1 4, 5 1104
          XM_011525095.1 XP_011523397.1 5, 6 1001
          XM_011525096.1 XP_011523398.1 3, 4 314
          XM_017024920.1 XP_016880409.1 3, 4 423
          GenBank mRNA AK098172.1 - 3, 4 523
          AK130192.1 - 4, 5 987
          AK292205.1 - 4, 5 1003
          AK303868.1 - 3, 4 812
          AK312564.1 - 4, 5 927
          AL834159.1 - 3, 4 805
          BC008727.2 - 4, 5 928 3163891
          BC071733.1 - 3, 4 747 5217484
          CR457438.1 - 3, 4 473
          HQ692826.1 - 3, 4 473
          HQ692827.1 - 3, 4 458
          S50916.1 - 335
          X06538.1 - 3, 4 699
          X06614.1 - 3, 4 575

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          C4 zinc finger nuclear receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          ureteric bud development
          neural tube closure
          liver development
          glandular epithelial cell development
          outflow tract septum morphogenesis
          growth plate cartilage development
          transcription initiation from RNA polymerase II promoter
          protein phosphorylation
          signal transduction
          germ cell development
          spermatogenesis
          female pregnancy
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          hippocampus development
          prostate gland development
          negative regulation of granulocyte differentiation
          embryonic camera-type eye development
          regulation of myelination
          response to estradiol
          response to retinoic acid
          negative regulation of interferon-gamma production
          negative regulation of tumor necrosis factor production
          positive regulation of interleukin-13 production
          positive regulation of interleukin-4 production
          positive regulation of interleukin-5 production
          response to vitamin A
          response to cytokine
          multicellular organism growth
          negative regulation of apoptotic process
          apoptotic cell clearance
          steroid hormone mediated signaling pathway
          response to ethanol
          positive regulation of T-helper 2 cell differentiation
          positive regulation of neuron differentiation
          positive regulation of cell cycle
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of translational initiation
          regulation of synaptic plasticity
          retinoic acid receptor signaling pathway
          positive regulation of binding
          ventricular cardiac muscle cell differentiation
          Sertoli cell fate commitment
          limb development
          face development
          trachea cartilage development
          chondroblast differentiation
          negative regulation of cartilage development
          cellular response to lipopolysaccharide
          cellular response to retinoic acid
          cellular response to estrogen stimulus
          translation repressor activity, nucleic acid binding
          RNA polymerase II regulatory region sequence-specific DNA binding
          retinoic acid binding
          transcription factor activity, sequence-specific DNA binding
          steroid hormone receptor activity
          retinoic acid receptor activity
          transcription coactivator activity
          transcription corepressor activity
          receptor binding
          protein binding
          transcription factor binding
          drug binding
          zinc ion binding
          enzyme binding
          protein domain specific binding
          chromatin DNA binding
          histone deacetylase binding
          protein kinase B binding
          retinoic acid-responsive element binding
          protein heterodimerization activity
          mRNA 5'-UTR binding
          protein kinase A binding
          alpha-actinin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0