Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RPL6 SIRNA Assays ›
          Gene Symbol:
          RPL6 RPL6P27
          Gene Name:
          ribosomal protein L6
          ribosomal protein L6 pseudogene 27
          Gene Aliases:
          L6, SHUJUN-2, TAXREB107, TXREB1
          RPL6_10_1582
          Chromosome Location:
          Chr.12: 112405181 - 112418850 on Build GRCh38
          Chromosome Location:
          Chr.18: 6462110 - 6463029 on Build GRCh38
          UniGene ID:
          Hs.528668
          Hs.689533
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 142170

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RPL6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000970.4 NP_000961.2 2, 3 286
          NM_001024662.2 NP_001019833.1 2, 3 462
          NM_001320137.1 NP_001307066.1 2, 3 467
          NM_001320138.1 NP_001307067.1 2, 3 501
          NM_001320139.1 NP_001307068.1 2, 3 348
          NM_001320140.1 NP_001307069.1 2, 3 510
          NM_001320141.1 NP_001307070.1 4, 5 575
          NM_001320142.1 NP_001307071.1 2, 3 222
          XM_017019781.1 XP_016875270.1 4, 5 1610
          XM_017019782.1 XP_016875271.1 4, 5 771
          GenBank mRNA AF261087.1 - 1 253
          AK298117.1 - 2, 3 248
          AK299470.1 - 3, 4 216
          AK308469.1 - 2, 3 250
          AK311863.1 - 2, 3 250
          BC004138.2 - 1 262 2823733
          BC013863.1 - 1 236
          BC020679.1 - 1 250 4616668
          BC022444.1 - 2, 3 417 4286423
          BC031009.1 - 1 250 4333761
          BC032299.1 - 1 247 4994660
          BC071912.1 - 1 239 6293404
          BC104824.1 - 2, 3 262 8143827
          BC104826.1 - 2, 3 262 8143829
          BG030827.1 - 1 12
          BG722198.1 - 4, 5 576
          BP285025.1 - 2, 3 286
          BP308263.1 - 2, 3 467
          BU940020.1 - 285
          CB140996.1 - 2, 3 415
          CD049618.1 - 2, 3 432
          CD640506.1 - 2, 3 411
          D17554.1 - 2, 3 252
          HY016219.1 - 2, 3 296
          S71022.1 - 1, 2 96
          X69391.1 - 2, 3 257
          Gene Symbol: RPL6P27
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK126570.1 - 997

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          ribosomal large subunit assembly
          nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
          cytoplasmic translation
          regulation of transcription, DNA-templated
          rRNA processing
          translation
          translational initiation
          SRP-dependent cotranslational protein targeting to membrane
          viral transcription
          cell-cell adhesion
          DNA binding
          RNA binding
          structural constituent of ribosome
          poly(A) RNA binding
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0