Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PRKCZ SIRNA Assays ›
          Gene Symbol:
          PRKCZ
          Gene Name:
          protein kinase C zeta
          Gene Aliases:
          PKC-ZETA, PKC2
          Chromosome Location:
          Chr.1: 2050470 - 2185395 on Build GRCh38
          UniGene ID:
          Hs.496255
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 142298

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PRKCZ
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001033581.1 NP_001028753.1 5, 6 647
          NM_001033582.1 NP_001028754.1 5, 6 544
          NM_001242874.1 NP_001229803.1 5, 6 654
          NM_002744.4 NP_002735.3 8, 9 859
          XM_011541773.1 XP_011540075.1 8, 9 855
          XM_011541774.1 XP_011540076.1 5, 6 720
          XM_011541775.2 XP_011540077.1 8, 9 855
          XM_011541776.1 XP_011540078.1 5, 6 660
          XM_011541778.1 XP_011540080.1 5, 6 573
          XM_017001789.1 XP_016857278.1 8, 9 855
          XM_017001790.1 XP_016857279.1 8, 9 855
          XM_017001791.1 XP_016857280.1 5, 6 504
          XM_017001792.1 XP_016857281.1 5, 6 660
          XM_017001793.1 XP_016857282.1 5, 6 918
          XM_017001794.1 XP_016857283.1 5, 6 573
          XM_017001795.1 XP_016857284.1 5, 6 577
          XM_017001796.1 XP_016857285.1 5, 6 660
          XM_017001797.1 XP_016857286.1 3, 4 301
          XM_017001798.1 XP_016857287.1 5, 6 573
          XM_017001799.1 XP_016857288.1 5, 6 981
          XM_017001800.1 XP_016857289.1 3, 4 301
          XM_017001801.1 XP_016857290.1 3, 4 301
          XM_017001802.1 XP_016857291.1 5, 6 857
          XM_017001803.1 XP_016857292.1 8, 9 855
          XM_017001804.1 XP_016857293.1 5, 6 767
          XM_017001805.1 XP_016857294.1 5, 6 737
          GenBank mRNA AK097627.1 - 3, 4 280
          AK290995.1 - 8, 9 698
          AK294649.1 - 5, 6 649
          AK294782.1 - 5, 6 567
          AK302160.1 - 3, 4 271
          AK310372.1 - 3, 4 301
          AU134793.2 - 6 547
          BC008058.2 - 8, 9 806 2987996
          BC014270.2 - 8, 9 859 3835020
          BT007082.1 - 8, 9 698
          L14283.1 - 8, 9 698
          Z15108.1 - 8, 9 680

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule cytoskeleton organization
          positive regulation of cell-matrix adhesion
          protein phosphorylation
          inflammatory response
          signal transduction
          transforming growth factor beta receptor signaling pathway
          long-term memory
          positive regulation of cell proliferation
          insulin receptor signaling pathway
          cell migration
          peptidyl-serine phosphorylation
          establishment of cell polarity
          negative regulation of protein complex assembly
          actin cytoskeleton reorganization
          activation of phospholipase D activity
          activation of protein kinase B activity
          positive regulation of interleukin-4 production
          negative regulation of apoptotic process
          positive regulation of T-helper 2 cell differentiation
          positive regulation of glucose import
          negative regulation of insulin receptor signaling pathway
          positive regulation of insulin receptor signaling pathway
          vesicle transport along microtubule
          negative regulation of peptidyl-tyrosine phosphorylation
          positive regulation of NF-kappaB transcription factor activity
          protein heterooligomerization
          negative regulation of hydrolase activity
          membrane depolarization
          membrane hyperpolarization
          long-term synaptic potentiation
          positive regulation of ERK1 and ERK2 cascade
          protein kinase C signaling
          protein localization to plasma membrane
          neuron projection extension
          positive regulation of excitatory postsynaptic potential
          positive regulation of T-helper 2 cell cytokine production
          positive regulation of interleukin-5 secretion
          positive regulation of interleukin-13 secretion
          positive regulation of interleukin-10 secretion
          protein kinase activity
          protein serine/threonine kinase activity
          protein kinase C activity
          protein binding
          ATP binding
          potassium channel regulator activity
          protein kinase binding
          protein domain specific binding
          phospholipase binding
          insulin receptor substrate binding
          metal ion binding
          14-3-3 protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0