Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PSMD4 SIRNA Assays ›
          Gene Symbol:
          PSMD4 PIPSL
          Gene Name:
          proteasome 26S subunit, non-ATPase 4
          PIP5K1A and PSMD4-like, pseudogene
          Gene Aliases:
          AF, AF-1, ASF, MCB1, Rpn10, S5A, pUB-R5
          PIP5K1L1, PIP5K1P3, PSMD4P2, bA429H9.1
          Chromosome Location:
          Chr.1: 151254700 - 151267479 on Build GRCh38
          Chromosome Location:
          Chr.10: 93958140 - 93961915 on Build GRCh38
          UniGene ID:
          Hs.505059
          Hs.149255
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 142364

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PSMD4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002810.2 NP_002801.1 8 906
          XM_005245354.2 XP_005245411.1 8 936
          XM_006711464.3 XP_006711527.1 7 761
          XM_017001856.1 XP_016857345.1 7 747
          GenBank mRNA AB033605.1 - 7 1053
          BC002365.2 - 8 856 2820813
          BC072008.1 - 8 870 6285035
          U24704.1 - 8 906
          U72664.1 - 6 638
          Gene Symbol: PIPSL
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_002319.2 1 2818

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          protease

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          protein polyubiquitination
          stimulatory C-type lectin receptor signaling pathway
          antigen processing and presentation of exogenous peptide antigen via MHC class I, TAP-dependent
          regulation of cellular amino acid metabolic process
          anaphase-promoting complex-dependent catabolic process
          tumor necrosis factor-mediated signaling pathway
          NIK/NF-kappaB signaling
          Fc-epsilon receptor signaling pathway
          proteasome-mediated ubiquitin-dependent protein catabolic process
          proteasome assembly
          regulation of mRNA stability
          T cell receptor signaling pathway
          negative regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
          positive regulation of ubiquitin-protein ligase activity involved in regulation of mitotic cell cycle transition
          Wnt signaling pathway, planar cell polarity pathway
          negative regulation of canonical Wnt signaling pathway
          positive regulation of canonical Wnt signaling pathway
          phosphatidylinositol phosphorylation
          protein binding
          transcription factor binding
          polyubiquitin binding
          identical protein binding
          poly(A) RNA binding
          phosphatidylinositol phosphate kinase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0