Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SHC1 SIRNA Assays ›
Gene Symbol:
SHC1
Gene Name:
SHC adaptor protein 1
Gene Aliases:
SHC, SHCA
Chromosome Location:
Chr.1: 154962298 - 154974492 on Build GRCh38
UniGene ID:
Hs.433795
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
142622
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SHC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130040.1
NP_001123512.1
8, 9
1403
NM_001130041.1
NP_001123513.1
9, 10
1091
NM_001202859.1
NP_001189788.1
9, 10
1091
NM_003029.4
NP_003020.2
9, 10
1091
NM_183001.4
NP_892113.4
8, 9
1403
XM_005245449.4
XP_005245506.1
8, 9
2619
XM_005245451.4
XP_005245508.1
9, 10
1014
XM_011509892.2
XP_011508194.1
8, 9
2643
XM_011509893.2
XP_011508195.1
8, 9
2644
XM_011509894.2
XP_011508196.1
7, 8
2593
XM_011509897.1
XP_011508199.1
8, 9
1028
XM_011509898.2
XP_011508200.1
8, 9
932
XM_017002081.1
XP_016857570.1
7, 8
2561
XM_017002082.1
XP_016857571.1
7, 8
2562
XM_017002083.1
XP_016857572.1
8, 9
1001
GenBank mRNA
AB208849.1
-
5, 6
520
AB451255.1
-
8, 9
1182
AB451379.1
-
8, 9
1182
AK096169.1
-
8, 9
1386
AK130014.1
-
7, 8
736
AK225332.1
-
8, 9
1328
AK292143.1
-
9, 10
1091
AK296786.1
-
9, 10
1214
AK315842.1
-
9, 10
1002
BC014158.2
-
9, 10
972
4541216
BC033925.1
-
6, 7
805
4896385
BX647136.1
-
9, 10
1319
BX647149.1
-
8, 9
1403
U73377.1
-
8, 9
1376
X68148.1
-
9, 10
933
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
activation of MAPK activity
angiogenesis
response to hypoxia
epidermal growth factor receptor signaling pathway
regulation of epidermal growth factor-activated receptor activity
Ras protein signal transduction
heart development
aging
positive regulation of cell proliferation
insulin receptor signaling pathway
response to toxic substance
viral process
single organismal cell-cell adhesion
organ regeneration
neuron projection development
actin cytoskeleton reorganization
response to nicotine
IRE1-mediated unfolded protein response
Fc-epsilon receptor signaling pathway
ERBB2 signaling pathway
regulation of growth
response to hydrogen peroxide
positive regulation of GTPase activity
positive regulation of DNA replication
positive regulation of vasoconstriction
positive regulation of smooth muscle cell proliferation
leukocyte migration
response to glucocorticoid
cellular response to growth factor stimulus
phosphotyrosine binding
transmembrane receptor protein tyrosine kinase adaptor activity
Ras guanyl-nucleotide exchange factor activity
epidermal growth factor receptor binding
insulin receptor binding
insulin-like growth factor receptor binding
neurotrophin TRKA receptor binding
protein binding
phospholipid binding
ephrin receptor binding
protein phosphatase 2A binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0