Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PTK2 SIRNA Assays ›
Gene Symbol:
PTK2
Gene Name:
protein tyrosine kinase 2
Gene Aliases:
FADK, FAK, FAK1, FRNK, PPP1R71, p125FAK, pp125FAK
Chromosome Location:
Chr.8: 140658382 - 141001313 on Build GRCh38
UniGene ID:
Hs.395482
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
142860
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PTK2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001199649.1
NP_001186578.1
3
420
NM_001316342.1
NP_001303271.1
2
331
NM_005607.4
NP_005598.3
3
409
NM_153831.3
NP_722560.1
3
420
XM_005251003.2
XP_005251060.1
2
461
XM_005251012.2
XP_005251069.1
2
303
XM_005251013.2
XP_005251070.1
2
289
XM_006716608.2
XP_006716671.1
3
392
XM_006716609.2
XP_006716672.1
3
392
XM_006716610.2
XP_006716673.1
3
384
XM_011517194.2
XP_011515496.1
3
361
XM_017013651.1
XP_016869140.1
1
142
XM_017013652.1
XP_016869141.1
1
142
XM_017013653.1
XP_016869142.1
2
461
XM_017013654.1
XP_016869143.1
2
461
XM_017013655.1
XP_016869144.1
2
461
XM_017013656.1
XP_016869145.1
2
461
XM_017013657.1
XP_016869146.1
2
461
XM_017013658.1
XP_016869147.1
3
285
XM_017013659.1
XP_016869148.1
1
142
XM_017013660.1
XP_016869149.1
3
285
XM_017013661.1
XP_016869150.1
2
303
XM_017013662.1
XP_016869151.1
3
285
XM_017013663.1
XP_016869152.1
2
216
XM_017013664.1
XP_016869153.1
3
392
XM_017013665.1
XP_016869154.1
2
216
XM_017013666.1
XP_016869155.1
2
461
XM_017013667.1
XP_016869156.1
3
285
XM_017013668.1
XP_016869157.1
2
303
XM_017013669.1
XP_016869158.1
3
285
XM_017013670.1
XP_016869159.1
3
285
XM_017013671.1
XP_016869160.1
3
285
XM_017013672.1
XP_016869161.1
2
216
XM_017013673.1
XP_016869162.1
2
216
XM_017013674.1
XP_016869163.1
3
392
XM_017013675.1
XP_016869164.1
2
216
XM_017013676.1
XP_016869165.1
1
142
XM_017013677.1
XP_016869166.1
3
285
XM_017013678.1
XP_016869167.1
2
216
XM_017013679.1
XP_016869168.1
2
216
XM_017013681.1
XP_016869170.1
3
392
XM_017013682.1
XP_016869171.1
2
289
XM_017013683.1
XP_016869172.1
3
392
XM_017013684.1
XP_016869173.1
3
392
XM_017013685.1
XP_016869174.1
3
336
XM_017013686.1
XP_016869175.1
2
289
XM_017013687.1
XP_016869176.1
3
392
XM_017013688.1
XP_016869177.1
3
392
XM_017013689.1
XP_016869178.1
2
257
XM_017013690.1
XP_016869179.1
2
231
GenBank mRNA
AB209071.1
-
2
237
AK304356.1
-
3
265
AL832961.1
-
3
292
BC035404.2
-
1
141
4999680
BY800117.1
-
2
397
DA236314.1
-
3
392
DA994616.1
-
2
190
KF437463.1
-
1
113
L13616.1
-
3
342
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
microtubule cytoskeleton organization
angiogenesis
vasculogenesis
neuron migration
placenta development
regulation of protein phosphorylation
positive regulation of protein phosphorylation
heart morphogenesis
cellular component disassembly involved in execution phase of apoptosis
signal complex assembly
epidermal growth factor receptor signaling pathway
transforming growth factor beta receptor signaling pathway
integrin-mediated signaling pathway
axon guidance
positive regulation of cell proliferation
regulation of cell shape
embryo development
regulation of endothelial cell migration
regulation of epithelial cell migration
positive regulation of phosphatidylinositol 3-kinase signaling
peptidyl-tyrosine phosphorylation
central nervous system neuron axonogenesis
negative regulation of cell-cell adhesion
establishment of cell polarity
extracellular matrix organization
positive regulation of cell migration
regulation of cell adhesion mediated by integrin
netrin-activated signaling pathway
peptidyl-tyrosine autophosphorylation
Fc-gamma receptor signaling pathway involved in phagocytosis
establishment of nucleus localization
regulation of cell proliferation
negative regulation of apoptotic process
regulation of GTPase activity
endothelial cell migration
positive regulation of GTPase activity
positive regulation of phosphatidylinositol 3-kinase activity
innate immune response
regulation of osteoblast differentiation
positive regulation of protein kinase activity
negative regulation of organ growth
protein autophosphorylation
vascular endothelial growth factor receptor signaling pathway
ephrin receptor signaling pathway
cell motility
negative regulation of axonogenesis
regulation of cytoskeleton organization
regulation of focal adhesion assembly
positive regulation of protein kinase B signaling
negative regulation of synapse assembly
growth hormone receptor signaling pathway
regulation of substrate adhesion-dependent cell spreading
positive regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
negative regulation of anoikis
actin binding
protein kinase activity
protein tyrosine kinase activity
non-membrane spanning protein tyrosine kinase activity
signal transducer activity
Ras guanyl-nucleotide exchange factor activity
receptor binding
protein binding
ATP binding
JUN kinase binding
protein kinase binding
SH2 domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0