Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PRKDC SIRNA Assays ›
          Gene Symbol:
          PRKDC
          Gene Name:
          protein kinase, DNA-activated, catalytic polypeptide
          Gene Aliases:
          DNA-PKcs, DNAPK, DNPK1, HYRC, HYRC1, IMD26, XRCC7, p350
          Chromosome Location:
          Chr.8: 47773108 - 47960183 on Build GRCh38
          UniGene ID:
          Hs.491682
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 143011

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PRKDC
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001081640.1 NP_001075109.1 14 1520
          NM_006904.6 NP_008835.5 14 1520
          GenBank mRNA AK296833.1 - 14 1473
          U34994.3 - 14 1515
          U47077.5 - 14 1520

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          somitogenesis
          negative regulation of protein phosphorylation
          B cell lineage commitment
          pro-B cell differentiation
          T cell lineage commitment
          negative regulation of immunoglobulin production
          positive regulation of immune system process
          double-strand break repair
          double-strand break repair via nonhomologous end joining
          cellular protein modification process
          brain development
          heart development
          cell proliferation
          intrinsic apoptotic signaling pathway in response to DNA damage
          response to gamma radiation
          response to activity
          telomere capping
          peptidyl-serine phosphorylation
          protein destabilization
          positive regulation of type I interferon production
          cellular response to insulin stimulus
          T cell differentiation in thymus
          immunoglobulin V(D)J recombination
          T cell receptor V(D)J recombination
          ectopic germ cell programmed cell death
          regulation of circadian rhythm
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of fibroblast proliferation
          rhythmic process
          spleen development
          thymus development
          positive regulation of developmental growth
          regulation of smooth muscle cell proliferation
          signal transduction involved in mitotic G1 DNA damage checkpoint
          double-strand break repair via alternative nonhomologous end joining
          negative regulation of cellular senescence
          negative regulation of response to gamma radiation
          double-stranded DNA binding
          protein kinase activity
          protein serine/threonine kinase activity
          DNA-dependent protein kinase activity
          protein binding
          ATP binding
          transcription factor binding
          enzyme binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0