Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RORA SIRNA Assays ›
          Gene Symbol:
          RORA
          Gene Name:
          RAR related orphan receptor A
          Gene Aliases:
          NR1F1, ROR1, ROR2, ROR3, RZR-ALPHA, RZRA
          Chromosome Location:
          Chr.15: 60488284 - 61229303 on Build GRCh38
          UniGene ID:
          Hs.560343
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 143155

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RORA
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002943.3 NP_002934.1 11 1674
          NM_134260.2 NP_599022.1 12 1698
          NM_134261.2 NP_599023.1 11 1528
          NM_134262.2 NP_599024.1 10 1369
          XM_005254584.4 XP_005254641.1 11 5989
          XM_011521874.1 XP_011520176.1 10 1566
          XM_011521875.1 XP_011520177.1 11 1398
          XM_011521877.2 XP_011520179.1 10 1601
          XM_011521878.1 XP_011520180.1 12 1440
          XM_011521879.2 XP_011520181.1 8 1326
          XM_017022466.1 XP_016877955.1 10 3390
          XM_017022467.1 XP_016877956.1 10 3063
          GenBank mRNA AB307694.1 - 11 1443
          BC008831.2 - 11 1451 4121602
          BC100987.2 - 10 1319 40005784
          BC100988.1 - 10 1319 40005785
          BC100989.1 - 10 1319 40005786
          BC100990.2 - 10 1319 40005787
          HQ692818.1 - 10 1278
          HQ709174.1 - 10 1275
          L14611.1 - 10 1299
          U04897.1 - 11 1544
          U04898.1 - 12 1631
          U04899.1 - 11 1596

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          C4 zinc finger nuclear receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          regulation of transcription, DNA-templated
          transcription initiation from RNA polymerase II promoter
          xenobiotic metabolic process
          nitric oxide biosynthetic process
          regulation of smoothened signaling pathway
          positive regulation of vascular endothelial growth factor production
          regulation of glucose metabolic process
          regulation of steroid metabolic process
          cerebellar Purkinje cell differentiation
          cerebellar granule cell precursor proliferation
          intracellular receptor signaling pathway
          circadian regulation of gene expression
          cellular response to sterol
          muscle cell differentiation
          positive regulation of circadian rhythm
          regulation of macrophage activation
          negative regulation of I-kappaB kinase/NF-kappaB signaling
          steroid hormone mediated signaling pathway
          negative regulation of fat cell differentiation
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          cGMP metabolic process
          negative regulation of inflammatory response
          regulation of transcription involved in cell fate commitment
          triglyceride homeostasis
          cellular response to interleukin-1
          cellular response to tumor necrosis factor
          cellular response to hypoxia
          T-helper 17 cell differentiation
          regulation of cholesterol homeostasis
          RNA polymerase II regulatory region sequence-specific DNA binding
          core promoter sequence-specific DNA binding
          transcription corepressor binding
          transcription coactivator binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          steroid hormone receptor activity
          RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
          protein binding
          beta-catenin binding
          transcription factor binding
          oxysterol binding
          zinc ion binding
          sequence-specific DNA binding
          transcription factor activity, direct ligand regulated sequence-specific DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0