Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PFKM SIRNA Assays ›
          Gene Symbol:
          PFKM
          Gene Name:
          phosphofructokinase, muscle
          Gene Aliases:
          ATP-PFK, GSD7, PFK-1, PFK1, PFKA, PFKX, PPP1R122
          Chromosome Location:
          Chr.12: 48105278 - 48146404 on Build GRCh38
          UniGene ID:
          Hs.75160
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 143272

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PFKM
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000289.5 NP_000280.1 23 2579
          NM_001166686.1 NP_001160158.1 25 2881
          NM_001166687.1 NP_001160159.1 23 2433
          NM_001166688.1 NP_001160160.1 22 2522
          XM_005268974.1 XP_005269031.1 26 2778
          XM_005268975.1 XP_005269032.1 26 2917
          XM_005268976.3 XP_005269033.1 26 2960
          XM_005268977.1 XP_005269034.1 25 2764
          XM_005268978.2 XP_005269035.1 25 2677
          XM_005268979.1 XP_005269036.1 25 2764
          XM_011538487.1 XP_011536789.1 24 2619
          XM_011538488.2 XP_011536790.1 23 2544
          XM_017019467.1 XP_016874956.1 26 2847
          XM_017019468.1 XP_016874957.1 25 2699
          XM_017019469.1 XP_016874958.1 24 2671
          GenBank mRNA AK126020.1 - 27 3234
          AK126229.1 - 26 2831
          AK303681.1 - 18 1926
          AK308185.1 - 18 1902
          BC000534.2 - 22 2409 2964710
          BC007798.2 - 21 2296
          BC012799.2 - 22 2409 2964710
          BC013298.2 - 22 2409 2964710
          BC021203.2 - 23 2431 4100117
          BX537703.1 - 2 4285
          M26066.1 - 23 2470
          U24183.1 - 22 2722
          Y00698.1 - 22 2413

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          carbohydrate kinase kinase transferase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          glycogen catabolic process
          fructose 6-phosphate metabolic process
          glycolytic process
          positive regulation of insulin secretion
          glucose homeostasis
          muscle cell cellular homeostasis
          carbohydrate phosphorylation
          protein oligomerization
          canonical glycolysis
          glycolysis from storage polysaccharide through glucose-1-phosphate
          6-phosphofructokinase activity
          protein binding
          ATP binding
          protein C-terminus binding
          kinase binding
          identical protein binding
          protein homodimerization activity
          metal ion binding
          fructose binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0