Search
Search

LSP1| Gene Symbol: LSP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013253.1 | NP_001013271.1 | 2 | 371 | ||
| NM_001013254.1 | NP_001013272.1 | 2 | 601 | |||
| NM_001013255.1 | NP_001013273.1 | 2 | 107 | |||
| NM_001242932.1 | NP_001229861.1 | 3 | 602 | |||
| NM_001289005.1 | NP_001275934.1 | 2 | 467 | |||
| NM_002339.2 | NP_002330.1 | 2 | 285 | |||
| GenBank mRNA | AK056576.1 | - | 2 | 1226 | ||
| AK092071.1 | - | 3 | 602 | |||
| AK222733.1 | - | 2 | 181 | |||
| AK290279.1 | - | 2 | 123 | |||
| AK292033.1 | - | 2 | 469 | |||
| BC001785.1 | - | 2 | 177 | 3542307 | ||
| BQ893037.1 | - | 2 | 145 | |||
| BQ893821.1 | - | 107 | ||||
| CR541728.1 | - | 2 | 110 | |||
| CR541754.1 | - | 2 | 110 | |||
| S67783.1 | - | 2 | 110 | |||
| X55188.1 | - | 2 | 157 | |||