Search
Search

CNOT4| Gene Symbol: CNOT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008225.2 | NP_001008226.1 | 10 | 1754 | ||
| NM_001190847.1 | NP_001177776.1 | 10 | 1763 | |||
| NM_001190848.1 | NP_001177777.1 | 10 | 1763 | |||
| NM_001190849.1 | NP_001177778.1 | 10 | 1754 | |||
| NM_001190850.1 | NP_001177779.1 | 10 | 1763 | |||
| NM_013316.3 | NP_037448.2 | 10 | 1754 | |||
| XM_017012235.1 | XP_016867724.1 | 10 | 1624 | |||
| GenBank mRNA | AF091094.1 | - | 7 | 1028 | ||
| AK074671.1 | - | 7 | 990 | |||
| AK300365.1 | - | 10 | 1720 | |||
| AK314540.1 | - | 10 | 1719 | |||
| AL389980.1 | - | 10 | 1708 | |||
| BC035590.1 | - | 10 | 1714 | 5576434 | ||
| CA396599.1 | - | 1 | 24 | |||
| U71267.1 | - | 10 | 1713 | |||
| U71268.1 | - | 10 | 1713 | |||