Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KTN1 SIRNA Assays ›
Gene Symbol:
KTN1
Gene Name:
kinectin 1
Gene Aliases:
CG1, KNT, MU-RMS-40.19
Chromosome Location:
Chr.14: 55580207 - 55684584 on Build GRCh38
UniGene ID:
Hs.509414
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
144438
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KTN1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001079521.1
NP_001072989.1
2
567
NM_001079522.1
NP_001072990.1
3
691
NM_001271014.1
NP_001257943.1
3
691
NM_004986.3
NP_004977.2
2
567
NR_073128.1
3
691
NR_073129.1
3
691
XM_006720138.2
XP_006720201.1
3
682
XM_006720139.2
XP_006720202.1
2
530
XM_006720140.3
XP_006720203.1
2
530
XM_006720141.2
XP_006720204.1
2
530
XM_011536751.2
XP_011535053.1
2
788
XM_011536752.1
XP_011535054.1
2
530
XM_011536753.1
XP_011535055.1
2
530
XM_017021278.1
XP_016876767.1
2
6380
XM_017021279.1
XP_016876768.1
2
530
XM_017021280.1
XP_016876769.1
3
682
XM_017021281.1
XP_016876770.1
3
6504
XM_017021282.1
XP_016876771.1
3
685
XM_017021283.1
XP_016876772.1
3
619
XM_017021284.1
XP_016876773.1
2
530
XM_017021285.1
XP_016876774.1
3
662
XM_017021286.1
XP_016876775.1
2
530
GenBank mRNA
AK302797.1
-
3
645
AY264265.1
-
3
576
BC050555.1
-
2
506
BC112337.1
-
3
574
40068009
BC117132.1
-
2
508
40125683
BC143720.1
-
2
508
9052234
D13629.1
-
2
472
L25616.1
-
1
429
Z22551.1
-
2
458
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001293636, NM_008477, NM_001293635, L43326, BC046994, and others.
Gene Ontology Categories:
Function(s)
Process(es)
microtubule-based movement
cell-cell adhesion
protein binding
kinesin binding
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-xhq5n:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0