Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other KIT SIRNA Assays ›
          Gene Symbol:
          KIT
          Gene Name:
          KIT proto-oncogene receptor tyrosine kinase
          Gene Aliases:
          C-Kit, CD117, PBT, SCFR
          Chromosome Location:
          Chr.4: 54657928 - 54740715 on Build GRCh38
          UniGene ID:
          Hs.479754
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 144533

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: KIT
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000222.2 NP_000213.1 5 887
          NM_001093772.1 NP_001087241.1 5 887
          XM_005265740.1 XP_005265797.1 5 861
          XM_005265741.1 XP_005265798.1 5 861
          XM_005265742.2 XP_005265799.1 5 861
          XM_017008178.1 XP_016863667.1 5 858
          XM_017008179.1 XP_016863668.1 5 861
          XM_017008180.1 XP_016863669.1 5 858
          GenBank mRNA AK304031.1 - 5 858
          BC071593.1 - 5 876 4375615
          EU826594.1 - 5 800
          GU983671.1 - 5 805
          HM015525.1 - 5 805
          HM015526.1 - 5 805
          KT326922.1 - 5 818
          X06182.1 - 5 821

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          activation of MAPK activity
          ovarian follicle development
          myeloid progenitor cell differentiation
          lymphoid progenitor cell differentiation
          immature B cell differentiation
          dendritic cell cytokine production
          mast cell chemotaxis
          glycosphingolipid metabolic process
          inflammatory response
          signal transduction
          spermatogenesis
          spermatid development
          positive regulation of cell proliferation
          germ cell migration
          regulation of cell shape
          visual learning
          male gonad development
          positive regulation of gene expression
          positive regulation of phospholipase C activity
          regulation of phosphatidylinositol 3-kinase signaling
          positive regulation of phosphatidylinositol 3-kinase signaling
          peptidyl-tyrosine phosphorylation
          cytokine-mediated signaling pathway
          stem cell population maintenance
          lamellipodium assembly
          hemopoiesis
          T cell differentiation
          erythrocyte differentiation
          melanocyte differentiation
          positive regulation of cell migration
          positive regulation of pseudopodium assembly
          actin cytoskeleton reorganization
          mast cell cytokine production
          somatic stem cell population maintenance
          embryonic hemopoiesis
          ectopic germ cell programmed cell death
          hematopoietic stem cell migration
          megakaryocyte development
          Fc receptor signaling pathway
          Kit signaling pathway
          erythropoietin-mediated signaling pathway
          regulation of cell proliferation
          positive regulation of tyrosine phosphorylation of Stat1 protein
          positive regulation of tyrosine phosphorylation of Stat3 protein
          positive regulation of tyrosine phosphorylation of Stat5 protein
          negative regulation of programmed cell death
          mast cell degranulation
          positive regulation of MAPK cascade
          pigmentation
          positive regulation of GTPase activity
          positive regulation of phosphatidylinositol 3-kinase activity
          positive regulation of Notch signaling pathway
          positive regulation of JAK-STAT cascade
          protein autophosphorylation
          phosphatidylinositol phosphorylation
          phosphatidylinositol-mediated signaling
          regulation of developmental pigmentation
          somatic stem cell division
          positive regulation of long-term neuronal synaptic plasticity
          digestive tract development
          stem cell differentiation
          epithelial cell proliferation
          detection of mechanical stimulus involved in sensory perception of sound
          positive regulation of sequence-specific DNA binding transcription factor activity
          cell chemotaxis
          mast cell differentiation
          mast cell proliferation
          cellular response to thyroid hormone stimulus
          melanocyte migration
          melanocyte adhesion
          positive regulation of vascular smooth muscle cell differentiation
          protease binding
          protein tyrosine kinase activity
          transmembrane receptor protein tyrosine kinase activity
          receptor signaling protein tyrosine kinase activity
          stem cell factor receptor activity
          Ras guanyl-nucleotide exchange factor activity
          protein binding
          ATP binding
          cytokine binding
          protein homodimerization activity
          metal ion binding
          phosphatidylinositol-4,5-bisphosphate 3-kinase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0