Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GDNF SIRNA Assays ›
          Gene Symbol:
          GDNF
          Gene Name:
          glial cell derived neurotrophic factor
          Gene Aliases:
          ATF1, ATF2, HFB1-GDNF, HSCR3
          Chromosome Location:
          Chr.5: 37812677 - 37840044 on Build GRCh38
          UniGene ID:
          Hs.248114
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 144551

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: GDNF
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000514.3 NP_000505.1 3 790
          NM_001190468.1 NP_001177397.1 3 810
          NM_001190469.1 NP_001177398.1 3 732
          NM_001278098.1 NP_001265027.1 3 618
          NM_199231.2 NP_954701.1 3 635
          XM_011514030.2 XP_011512332.1 2 503
          XM_017009337.1 XP_016864826.1 3 1076
          GenBank mRNA AY052832.1 - 2 512
          BC069119.1 - 2 590 7216973
          BC069369.1 - 2 590 7262145
          BC128108.1 - 2 590 40112122
          BC128109.1 - 2 590 40112125
          CR541923.1 - 2 590
          DQ235474.1 - 3 592
          KU178093.1 - 2 590
          KU178094.1 - 2 512

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_001301332, NM_001301333, NM_010275, NM_001301357, BC119031, and others.

          Panther Classification:

          Molecular Function -

          neurotrophic factor

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          metanephros development
          branching involved in ureteric bud morphogenesis
          neural crest cell migration
          organ induction
          postsynaptic membrane organization
          mesenchymal to epithelial transition involved in metanephros morphogenesis
          signal transduction
          nervous system development
          positive regulation of cell proliferation
          adult locomotory behavior
          regulation of gene expression
          postganglionic parasympathetic fiber development
          peristalsis
          neuron projection development
          positive regulation of monooxygenase activity
          positive regulation of dopamine secretion
          negative regulation of apoptotic process
          negative regulation of neuron apoptotic process
          positive regulation of GTPase activity
          positive regulation of cell differentiation
          positive regulation of transcription from RNA polymerase II promoter
          mRNA stabilization
          enteric nervous system development
          sympathetic nervous system development
          regulation of dopamine uptake involved in synaptic transmission
          ureteric bud formation
          regulation of morphogenesis of a branching structure
          positive regulation of ureteric bud formation
          positive regulation of mesenchymal to epithelial transition involved in metanephros morphogenesis
          positive regulation of branching involved in ureteric bud morphogenesis
          regulation of stem cell differentiation
          negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
          Ras guanyl-nucleotide exchange factor activity
          receptor binding
          protein binding
          growth factor activity
          protein homodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0