Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CYR61 SIRNA Assays ›
          Gene Symbol:
          CYR61
          Gene Name:
          cysteine rich angiogenic inducer 61
          Gene Aliases:
          CCN1, GIG1, IGFBP10
          Chromosome Location:
          Chr.1: 85580761 - 85583967 on Build GRCh38
          UniGene ID:
          Hs.8867
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 144772

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CYR61
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001554.4 NP_001545.2 5 1115
          GenBank mRNA AF003114.1 - 4 763
          AF003594.1 - 5 1014
          AF031385.1 - 5 971
          AK096420.1 - 1 944
          AK223385.1 - 5 1116
          AK295430.1 - 5 1043
          AK314571.1 - 5 1116
          AL832891.1 - 1 762
          BC001271.1 - 5 1050 3454581
          BC009199.2 - 5 1099 3830296
          BC016952.1 - 5 1106 3886130
          CR536519.1 - 5 891
          U62015.1 - 5 1100
          Y11307.1 - 5 1114
          Y12084.1 - 5 1105
          Z98053.1 - 5 1109

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          growth factor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cell growth
          osteoblast differentiation
          positive regulation of protein phosphorylation
          intussusceptive angiogenesis
          atrioventricular valve morphogenesis
          apoptotic process involved in heart morphogenesis
          ventricular septum development
          chemotaxis
          cell adhesion
          cell-cell signaling
          cell proliferation
          anatomical structure morphogenesis
          positive regulation of phospholipase activity
          positive regulation of cell-substrate adhesion
          extracellular matrix organization
          positive regulation of cell migration
          positive regulation of BMP signaling pathway
          positive regulation of osteoblast proliferation
          negative regulation of apoptotic process
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          wound healing, spreading of cells
          positive regulation of osteoblast differentiation
          positive regulation of protein kinase activity
          positive regulation of transcription from RNA polymerase II promoter
          atrial septum morphogenesis
          negative regulation of cell death
          chondroblast differentiation
          chorio-allantoic fusion
          labyrinthine layer blood vessel development
          positive regulation of cartilage development
          regulation of ERK1 and ERK2 cascade
          reactive oxygen species metabolic process
          positive regulation of ceramide biosynthetic process
          integrin binding
          insulin-like growth factor binding
          heparin binding
          extracellular matrix binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0