Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GLUL SIRNA Assays ›
          Gene Symbol:
          GLUL SBF2
          Gene Name:
          glutamate-ammonia ligase
          SET binding factor 2
          Gene Aliases:
          GLNS, GS, PIG43, PIG59
          CMT4B2, DENND7B, MTMR13
          Chromosome Location:
          Chr.1: 182378093 - 182392206 on Build GRCh38
          Chromosome Location:
          Chr.11: 9778667 - 10294216 on Build GRCh38
          UniGene ID:
          Hs.132016
          Hs.577252
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 144849

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: GLUL
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001033044.3 NP_001028216.1 4 890
          NM_001033056.3 NP_001028228.1 4 592
          NM_002065.6 NP_002056.2 5 1246
          XM_006711278.1 XP_006711341.1 4 953
          GenBank mRNA AK122784.1 - 4 890
          AL161952.1 - 2 1507
          AY486122.1 - 3 349
          AY513283.1 - 4 405
          AY947343.1 - 4 450
          BC010037.2 - 4 462 3545927
          BC011700.2 - 4 458 3528830
          BC011852.2 - 4 469 4137547
          BC018992.1 - 5 809 3352740
          BC031964.1 - 4 992 4837475
          BC051726.1 - 4 1655 5261804
          BC127883.1 - 3 260
          BG771732.1 - 4 509
          BX537384.1 - 4 474
          CD688156.1 - 4 583
          DA151535.1 - 4 535
          S70290.1 - 3 349
          X59834.1 - 4 458
          Y00387.2 - 4 468
          Gene Symbol: SBF2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AL832942.1 - 3 2771

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ligase phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          neurotransmitter uptake
          glutamate catabolic process
          glutamine biosynthetic process
          cell proliferation
          cellular amino acid biosynthetic process
          cellular response to starvation
          response to glucose
          ammonia assimilation cycle
          positive regulation of insulin secretion
          positive regulation of epithelial cell proliferation
          protein homooligomerization
          positive regulation of synaptic transmission, glutamatergic
          magnesium ion binding
          glutamate decarboxylase activity
          glutamate-ammonia ligase activity
          protein binding
          ATP binding
          glutamate binding
          manganese ion binding
          identical protein binding
          dynein light chain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0