Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GRB2 SIRNA Assays ›
Gene Symbol:
GRB2
Gene Name:
growth factor receptor bound protein 2
Gene Aliases:
ASH, EGFRBP-GRB2, Grb3-3, MST084, MSTP084, NCKAP2
Chromosome Location:
Chr.17: 75318076 - 75405709 on Build GRCh38
UniGene ID:
Hs.444356
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
144867
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GRB2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_002086.4
NP_002077.1
6
1997
NM_203506.2
NP_987102.1
5
1875
GenBank mRNA
BQ962129.1
-
1
408
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
epidermal growth factor receptor signaling pathway
Ras protein signal transduction
cell-cell signaling
axon guidance
insulin receptor signaling pathway
fibroblast growth factor receptor signaling pathway
positive regulation of signal transduction
regulation of phosphatidylinositol 3-kinase signaling
viral process
positive regulation of actin filament polymerization
T cell costimulation
receptor internalization
phosphatidylinositol-3-phosphate biosynthetic process
Fc-epsilon receptor signaling pathway
Fc-gamma receptor signaling pathway involved in phagocytosis
ERBB2 signaling pathway
negative regulation of epidermal growth factor receptor signaling pathway
signal transduction in response to DNA damage
regulation of MAPK cascade
positive regulation of GTPase activity
phosphatidylinositol phosphorylation
phosphatidylinositol-mediated signaling
anatomical structure formation involved in morphogenesis
leukocyte migration
branching involved in labyrinthine layer morphogenesis
cellular response to ionizing radiation
positive regulation of reactive oxygen species metabolic process
SH3/SH2 adaptor activity
Ras guanyl-nucleotide exchange factor activity
epidermal growth factor receptor binding
neurotrophin TRKA receptor binding
protein binding
1-phosphatidylinositol-3-kinase activity
SH3 domain binding
protein kinase binding
protein phosphatase binding
identical protein binding
insulin receptor substrate binding
poly(A) RNA binding
ephrin receptor binding
phosphatidylinositol-4,5-bisphosphate 3-kinase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0