Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ILK SIRNA Assays ›
          Gene Symbol:
          ILK
          Gene Name:
          integrin linked kinase
          Gene Aliases:
          HEL-S-28, ILK-1, ILK-2, P59, p59ILK
          Chromosome Location:
          Chr.11: 6603708 - 6610874 on Build GRCh38
          UniGene ID:
          Hs.706355
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 145118

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ILK
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001014794.2 NP_001014794.1 3 406
          NM_001014795.2 NP_001014795.1 2 707
          NM_001278441.1 NP_001265370.1 3 452
          NM_004517.3 NP_004508.1 3 452
          XM_005252904.4 XP_005252961.1 3 413
          XM_011520065.1 XP_011518367.1 3 412
          XM_017017672.1 XP_016873161.1 3 391
          GenBank mRNA AB209416.1 - 1 4388
          AK296628.1 - 3 373
          AK314843.1 - 3 387
          BC001554.1 - 3 313 3457801
          CB113885.1 - 3 429
          CR407673.1 - 2 245
          CR749220.1 - 2 701
          EU794625.1 - 2 540
          U40282.1 - 3 401

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          branching involved in ureteric bud morphogenesis
          positive regulation of cell-matrix adhesion
          outflow tract morphogenesis
          protein phosphorylation
          negative regulation of protein kinase activity
          cell cycle arrest
          cell-matrix adhesion
          integrin-mediated signaling pathway
          cell aging
          cell proliferation
          positive regulation of cell proliferation
          negative regulation of cardiac muscle cell apoptotic process
          fibroblast migration
          negative regulation of smooth muscle cell migration
          peptidyl-serine phosphorylation
          nerve development
          myelination in peripheral nervous system
          positive regulation of cell migration
          positive regulation of BMP signaling pathway
          myelin assembly
          regulation of actin cytoskeleton organization
          cell junction assembly
          substrate adhesion-dependent cell spreading
          positive regulation of phosphorylation
          extracellular fibril organization
          positive regulation of MAP kinase activity
          protein kinase B signaling
          negative regulation of neuron apoptotic process
          establishment or maintenance of epithelial cell apical/basal polarity
          positive regulation of myoblast differentiation
          positive regulation of osteoblast differentiation
          positive regulation of axon extension
          positive regulation of transcription, DNA-templated
          negative regulation of smooth muscle cell proliferation
          neuron projection morphogenesis
          positive regulation of dendrite morphogenesis
          protein heterooligomerization
          positive regulation of protein kinase B signaling
          platelet aggregation
          positive regulation of canonical Wnt signaling pathway
          negative regulation of neural precursor cell proliferation
          protein serine/threonine kinase activity
          signal transducer activity
          integrin binding
          protein binding
          ATP binding
          SH3 domain binding
          protein kinase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-s9xdw:80/100.66.76.163:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0