Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other INPP5D SIRNA Assays ›
          Gene Symbol:
          INPP5D
          Gene Name:
          inositol polyphosphate-5-phosphatase D
          Gene Aliases:
          SHIP, SHIP-1, SHIP1, SIP-145, hp51CN, p150Ship
          Chromosome Location:
          Chr.2: 233059967 - 233207903 on Build GRCh38
          UniGene ID:
          Hs.262886
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 145278

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: INPP5D
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001017915.2 NP_001017915.1 19 2601
          NM_005541.4 NP_005532.2 19 2598
          XM_017004004.1 XP_016859493.1 19 2177
          GenBank mRNA BC099920.1 - 19 2209 6304992
          BC113580.1 - 19 2146 8322632
          BC113582.1 - 19 2146 8322634
          U50040.1 - 14 1466
          U50041.1 - 19 2210
          U53470.1 - 19 2219
          U57650.1 - 19 2598
          U84400.1 - 19 2170
          X98429.1 - 19 2226

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          phosphatidylinositol biosynthetic process
          phosphate-containing compound metabolic process
          apoptotic process
          signal transduction
          determination of adult lifespan
          negative regulation of signal transduction
          immunoglobulin mediated immune response
          negative regulation of B cell proliferation
          intracellular signal transduction
          positive regulation of apoptotic process
          inositol phosphate metabolic process
          negative regulation of interleukin-6 biosynthetic process
          positive regulation of B cell differentiation
          positive regulation of erythrocyte differentiation
          negative regulation of monocyte differentiation
          negative regulation of neutrophil differentiation
          negative regulation of osteoclast differentiation
          negative regulation of bone resorption
          phosphatidylinositol dephosphorylation
          negative regulation of immune response
          T cell receptor signaling pathway
          leukocyte migration
          inositol-polyphosphate 5-phosphatase activity
          protein binding
          phosphatidylinositol-3,4,5-trisphosphate 3-phosphatase activity
          SH3 domain binding
          phosphatidylinositol-3,4,5-trisphosphate 5-phosphatase activity
          PTB domain binding
          inositol-1,3,4,5-tetrakisphosphate 5-phosphatase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0