Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GATA2 SIRNA Assays ›
          Gene Symbol:
          GATA2
          Gene Name:
          GATA binding protein 2
          Gene Aliases:
          DCML, IMD21, MONOMAC, NFE1B
          Chromosome Location:
          Chr.3: 128479422 - 128493187 on Build GRCh38
          UniGene ID:
          Hs.367725
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 145417

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: GATA2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001145661.1 NP_001139133.1 4, 5 1298
          NM_001145662.1 NP_001139134.1 3, 4 1119
          NM_032638.4 NP_116027.2 3, 4 1197
          GenBank mRNA AK314826.1 - 4, 5 1298
          BC002557.2 - 3, 4 1197 3139044
          BC015577.2 - 3, 4 1114 4127146
          BC015613.2 - 3, 4 944 4811349
          BC018988.2 - 3, 4 1114 4128414
          BC051272.1 - 3, 4 1114 4893358
          BC051342.1 - 3, 4 1195 6341380
          BT006671.1 - 2, 3 863
          BX647499.1 - 2, 3 318
          M68891.1 - 3, 4 1143

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          urogenital system development
          cell fate determination
          neuron migration
          embryonic placenta development
          transcription, DNA-templated
          transcription from RNA polymerase II promoter
          phagocytosis
          positive regulation of cytosolic calcium ion concentration
          heart development
          blood coagulation
          organ morphogenesis
          tissue development
          regulation of primitive erythrocyte differentiation
          ventral spinal cord interneuron differentiation
          cell differentiation in hindbrain
          commitment of neuronal cell to specific neuron type in forebrain
          central nervous system neuron development
          pituitary gland development
          response to lipid
          somatic stem cell population maintenance
          regulation of histone acetylation
          eosinophil fate commitment
          inner ear morphogenesis
          mast cell degranulation
          negative regulation of fat cell differentiation
          positive regulation of erythrocyte differentiation
          negative regulation of macrophage differentiation
          positive regulation of megakaryocyte differentiation
          positive regulation of neuron differentiation
          negative regulation of Notch signaling pathway
          positive regulation of angiogenesis
          positive regulation of transcription from RNA polymerase II promoter
          cell maturation
          digestive tract development
          homeostasis of number of cells within a tissue
          positive regulation of phagocytosis
          positive regulation of phagocytosis, engulfment
          definitive hemopoiesis
          semicircular canal development
          negative regulation of fat cell proliferation
          GABAergic neuron differentiation
          negative regulation of neural precursor cell proliferation
          regulation of forebrain neuron differentiation
          RNA polymerase II regulatory region sequence-specific DNA binding
          RNA polymerase II core promoter sequence-specific DNA binding
          RNA polymerase II distal enhancer sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          RNA polymerase II transcription factor binding
          enhancer sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          transcription factor binding
          zinc ion binding
          C2H2 zinc finger domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0