Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FGF14 SIRNA Assays ›
Gene Symbol:
FGF14
Gene Name:
fibroblast growth factor 14
Gene Aliases:
FGF-14, FHF-4, FHF4, SCA27
Chromosome Location:
Chr.13: 101720855 - 102402428 on Build GRCh38
UniGene ID:
Hs.508616
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
145453
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FGF14
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001321931.1
NP_001308860.1
3, 4
565
NM_001321932.1
NP_001308861.1
3, 4
651
NM_001321933.1
NP_001308862.1
3, 4
561
NM_001321934.1
NP_001308863.1
2, 3
324
NM_001321935.1
NP_001308864.1
2, 3
177
NM_001321936.1
NP_001308865.1
3, 4
504
NM_001321937.1
NP_001308866.1
2, 3
670
NM_001321938.1
NP_001308867.1
4, 5
552
NM_001321940.1
NP_001308869.1
3, 4
907
NM_001321941.1
NP_001308870.1
3, 4
739
NM_001321942.1
NP_001308871.1
3, 4
746
NM_001321943.1
NP_001308872.1
3, 4
783
NM_001321944.1
NP_001308873.1
3, 4
999
NM_001321945.1
NP_001308874.1
3, 4
542
NM_001321946.1
NP_001308875.1
4, 5
648
NM_001321947.1
NP_001308876.1
3, 4
466
NM_001321948.1
NP_001308877.1
3, 4
538
NM_001321949.1
NP_001308878.1
3, 4
1235
NM_004115.3
NP_004106.1
2, 3
301
NM_175929.2
NP_787125.1
2, 3
412
GenBank mRNA
AY188178.1
-
2, 3
412
BC100920.1
-
2, 3
411
40003901
BC100921.1
-
2, 3
411
40003902
BC100922.2
-
2, 3
411
40003903
U66200.1
-
2, 3
301
Back To Top
More Information
Panther Classification:
Molecular Function -
growth factor
intercellular signal molecule
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of defense response to virus by host
signal transduction
cell-cell signaling
nervous system development
regulation of synaptic plasticity
regulation of postsynaptic membrane potential
xenophagy
positive regulation of high voltage-gated calcium channel activity
regulation of synaptic vesicle recycling
protein binding
growth factor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0