Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ESR1 SIRNA Assays ›
          Gene Symbol:
          ESR1
          Gene Name:
          estrogen receptor 1
          Gene Aliases:
          ER, ESR, ESRA, ESTRR, Era, NR3A1
          Chromosome Location:
          Chr.6: 151654148 - 152103274 on Build GRCh38
          UniGene ID:
          Hs.208124
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 145538

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ESR1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000125.3 NP_000116.2 3, 4 999
          NM_001122740.1 NP_001116212.1 4, 5 1026
          NM_001122741.1 NP_001116213.1 4, 5 983
          NM_001122742.1 NP_001116214.1 5, 6 1135
          NM_001291230.1 NP_001278159.1 4, 5 989
          NM_001328100.1 NP_001315029.1 3, 4 478
          XM_006715374.3 XP_006715437.1 5, 6 1135
          XM_006715375.3 XP_006715438.1 3, 4 1132
          XM_011535543.2 XP_011533845.1 6, 7 2960
          XM_011535544.2 XP_011533846.1 4, 5 1144
          XM_011535545.2 XP_011533847.1 4, 5 1033
          XM_011535547.2 XP_011533849.1 5, 6 1135
          XM_017010376.1 XP_016865865.1 5, 6 3582
          XM_017010377.1 XP_016865866.1 6, 7 3641
          XM_017010378.1 XP_016865867.1 5, 6 1093
          XM_017010379.1 XP_016865868.1 5, 6 1127
          XM_017010380.1 XP_016865869.1 4, 5 3451
          XM_017010381.1 XP_016865870.1 4, 5 879
          XM_017010382.1 XP_016865871.1 2, 3 199
          GenBank mRNA AB307713.1 - 3, 4 765
          AK304833.1 - 4, 5 806
          AK309580.1 - 5, 6 1033
          AK313644.1 - 4, 5 957
          BC128573.1 - 3, 4 870 40128594
          BC128574.1 - 3, 4 870 40128595
          BX640939.1 - 3, 4 479
          EU930361.1 - 2, 3 174
          EU982838.1 - 2, 3 194
          FJ011557.1 - 2, 3 186
          FJ147552.1 - 2, 3 194
          FJ147554.1 - 2, 3 194
          FJ147557.1 - 2, 3 186
          FJ147558.1 - 2, 3 192
          FJ169503.1 - 2, 3 186
          FJ716811.1 - 186
          FJ716812.1 - 2 186
          FJ716814.1 - 1, 2 66
          FJ716815.1 - 2, 3 186
          GQ411253.1 - 186
          GQ438791.1 - 1, 2 27
          JF810888.1 - 3, 4 765
          LC120323.1 - 3, 4 835
          LC120324.1 - 3, 4 835
          LC120325.1 - 3, 4 835
          LC120326.1 - 3, 4 835
          LC120327.1 - 3, 4 835
          LC120328.1 - 3, 4 835
          LC120329.1 - 3, 4 835
          LC120330.1 - 3, 4 835
          M12674.1 - 3, 4 1057
          U47678.1 - 765
          X03635.1 - 3, 4 1125

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          antral ovarian follicle growth
          osteoblast development
          chromatin remodeling
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          transcription initiation from RNA polymerase II promoter
          signal transduction
          phospholipase C-activating G-protein coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          androgen metabolic process
          negative regulation of gene expression
          positive regulation of phospholipase C activity
          intracellular steroid hormone receptor signaling pathway
          intracellular estrogen receptor signaling pathway
          response to estradiol
          negative regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation of sequence-specific DNA binding transcription factor activity
          regulation of neuron apoptotic process
          response to estrogen
          positive regulation of epidermal growth factor receptor signaling pathway
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of glucose import
          decidualization
          positive regulation of fibroblast proliferation
          negative regulation of smooth muscle cell proliferation
          positive regulation of epithelial cell proliferation
          positive regulation of sequence-specific DNA binding transcription factor activity
          Sertoli cell development
          Sertoli cell proliferation
          uterus development
          vagina development
          prostate epithelial cord elongation
          prostate epithelial cord arborization involved in prostate glandular acinus morphogenesis
          regulation of branching involved in prostate gland morphogenesis
          mammary gland branching involved in pregnancy
          mammary gland alveolus development
          epithelial cell proliferation involved in mammary gland duct elongation
          positive regulation of ERK1 and ERK2 cascade
          protein localization to chromatin
          cellular response to estrogen stimulus
          cellular response to estradiol stimulus
          negative regulation of triglyceride metabolic process
          negative regulation of neuron death
          negative regulation of production of miRNAs involved in gene silencing by miRNA
          baculum development
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          core promoter sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          transcription coactivator binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          steroid hormone receptor activity
          steroid binding
          protein binding
          beta-catenin binding
          transcription factor binding
          zinc ion binding
          enzyme binding
          nitric-oxide synthase regulator activity
          estrogen receptor activity
          type 1 metabotropic glutamate receptor binding
          protein complex binding
          estrogen response element binding
          phosphatidylinositol 3-kinase regulatory subunit binding
          RNA polymerase II transcription factor activity, estrogen-activated sequence-specific DNA binding
          hormone binding
          identical protein binding
          ATPase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-89x45:80/100.66.77.245:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0