Search
Search

F8| Gene Symbol: F8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000132.3 | NP_000123.1 | 23 | 6659 | ||
| NM_019863.2 | NP_063916.1 | 2 | 228 | |||
| GenBank mRNA | AK289947.1 | - | 2 | 158 | ||
| AK295164.1 | - | 11 | 1456 | |||
| AK313707.1 | - | 2 | 113 | |||
| AX746360.1 | - | 23 | 6488 | |||
| BC022513.1 | - | 2 | 147 | |||
| BC064380.1 | - | 2 | 207 | |||
| BC098389.1 | - | 2 | 226 | |||
| BC111967.1 | - | 2 | 116 | |||
| BC111969.1 | - | 2 | 115 | |||
| K01740.1 | - | 23 | 6638 | |||
| M14113.1 | - | 23 | 6659 | |||
| M90707.1 | - | 2 | 123 | |||
| X01179.1 | - | 23 | 6597 | |||