Search
Search

F8| Gene Symbol: F8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000132.3 | NP_000123.1 | 26 | 8531 | ||
| NM_019863.2 | NP_063916.1 | 5 | 2100 | |||
| GenBank mRNA | BC022513.1 | - | 5 | 2019 | ||
| BC064380.1 | - | 5 | 2079 | |||
| BC098389.1 | - | 5 | 2098 | |||
| K01740.1 | - | 26 | 8510 | |||
| M14113.1 | - | 26 | 8531 | |||
| M90707.1 | - | 5 | 1995 | |||
| X01179.1 | - | 26 | 8469 | |||