Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MTRF1 SIRNA Assays ›
Gene Symbol:
MTRF1
Gene Name:
mitochondrial translational release factor 1
Gene Aliases:
MRF1, MTTRF1, RF1
Chromosome Location:
Chr.13: 41216369 - 41311794 on Build GRCh38
UniGene ID:
Hs.382176
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
14563
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MTRF1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_004294.2
NP_004285.2
7
1043
XM_005266599.2
XP_005266656.1
9
1292
XM_006719897.2
XP_006719960.1
9
1400
XM_006719898.2
XP_006719961.1
9
1384
XM_006719900.2
XP_006719963.1
8
1113
XM_011535315.2
XP_011533617.1
9
1509
XM_011535317.2
XP_011533619.1
8
1392
XM_011535318.2
XP_011533620.1
9
1552
XM_011535320.2
XP_011533622.1
9
1488
XM_011535321.2
XP_011533623.1
12
1624
XM_011535323.2
XP_011533625.1
9
1354
XM_011535325.2
XP_011533627.1
8
1307
XM_011535326.2
XP_011533628.1
8
1353
XM_011535327.2
XP_011533629.1
7
1180
XM_017020856.1
XP_016876345.1
8
1353
XM_017020857.1
XP_016876346.1
11
1487
XM_017020858.1
XP_016876347.1
10
1482
XM_017020859.1
XP_016876348.1
8
1353
XM_017020860.1
XP_016876349.1
10
1370
XM_017020861.1
XP_016876350.1
10
1407
XM_017020862.1
XP_016876351.1
9
2565
XM_017020863.1
XP_016876352.1
9
2413
XM_017020864.1
XP_016876353.1
9
2305
XM_017020865.1
XP_016876354.1
8
2257
XM_017020866.1
XP_016876355.1
7
2122
XM_017020867.1
XP_016876356.1
9
1270
XM_017020868.1
XP_016876357.1
7
1238
XM_017020869.1
XP_016876358.1
8
1136
XM_017020870.1
XP_016876359.1
9
1400
XM_017020871.1
XP_016876360.1
8
1113
XM_017020872.1
XP_016876361.1
8
1353
XM_017020873.1
XP_016876362.1
12
1694
XM_017020874.1
XP_016876363.1
9
1246
XM_017020875.1
XP_016876364.1
9
1240
XM_017020876.1
XP_016876365.1
8
2580
XM_017020877.1
XP_016876366.1
10
1415
XM_017020878.1
XP_016876367.1
9
1292
XM_017020879.1
XP_016876368.1
7
2192
GenBank mRNA
AF072934.1
-
7
1014
AK055086.1
-
9
1414
AK294343.1
-
8
1200
BC042196.1
-
7
1043
5198308
Back To Top
More Information
Panther Classification:
Molecular Function -
translation release factor
Gene Ontology Categories:
Function(s)
Process(es)
regulation of translational termination
mitochondrial translational termination
translation release factor activity
translation release factor activity, codon specific
ribosome binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0