Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TMX2-CTNND1 SIRNA Assays ›
Gene Symbol:
TMX2-CTNND1
CTNND1
Gene Name:
TMX2-CTNND1 readthrough (NMD candidate)
catenin delta 1
Gene Aliases:
-
CAS, CTNND, P120CAS, P120CTN, p120, p120(CAS), p120(CTN)
Chromosome Location:
Chr.11: 57712523 - 57819180 on Build GRCh38
Chromosome Location:
Chr.11: 57761762 - 57819180 on Build GRCh38
UniGene ID:
Hs.166011
Hs.166011
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
145776
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TMX2-CTNND1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NR_037646.1
8
1968
GenBank mRNA
AB002382.1
-
8
1874
Gene Symbol: CTNND1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001085458.1
NP_001078927.1
7
1980
NM_001085459.1
NP_001078928.1
7
1980
NM_001085460.1
NP_001078929.1
7
1907
NM_001085461.1
NP_001078930.1
7
1980
NM_001085462.1
NP_001078931.1
6
1788
NM_001085463.1
NP_001078932.1
5
1499
NM_001085464.1
NP_001078933.1
5
1499
NM_001085465.1
NP_001078934.1
5
1499
NM_001085466.1
NP_001078935.1
5
1499
NM_001085467.1
NP_001078936.1
5
1572
NM_001085468.1
NP_001078937.1
5
1499
NM_001085469.1
NP_001078938.1
4
1427
NM_001206883.1
NP_001193812.1
6
1679
NM_001206884.1
NP_001193813.1
6
1679
NM_001206885.1
NP_001193814.1
7
1980
NM_001206886.1
NP_001193815.1
6
1679
NM_001206887.1
NP_001193816.1
6
1679
NM_001206888.1
NP_001193817.1
6
1679
NM_001206889.1
NP_001193818.1
7
1798
NM_001206890.1
NP_001193819.1
5
1521
NM_001206891.1
NP_001193820.1
7
1871
NM_001331.2
NP_001322.1
7
1980
GenBank mRNA
AF062317.1
-
7
1945
AF062318.1
-
2
703
AF062319.1
-
6
1644
AF062320.1
-
6
1644
AF062321.1
-
7
1945
AF062322.1
-
6
1644
AF062323.1
-
5
1464
AF062324.1
-
7
1872
AF062325.1
-
6
1644
AF062326.1
-
2
703
AF062327.1
-
6
1644
AF062328.1
-
7
1945
AF062329.1
-
7
1763
AF062330.1
-
5
1486
AF062331.1
-
7
1836
AF062332.1
-
5
1464
AF062333.1
-
2
703
AF062334.1
-
5
1537
AF062335.1
-
5
1464
AF062336.1
-
4
1392
AF062337.1
-
5
1464
AF062338.1
-
5
1464
AF062339.1
-
7
1945
AF062340.1
-
2
776
AF062341.1
-
7
1945
AF062342.1
-
2
703
AF062343.1
-
6
1753
AF062344.1
-
2
703
AK292554.1
-
7
1922
BC010501.1
-
3
1101
BC075795.1
-
7
1796
5269008
BT007333.1
-
1
440
CR749275.1
-
7
1740
Back To Top
More Information
Panther Classification:
Molecular Function -
intermediate filament binding protein
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
cell adhesion
brain development
Wnt signaling pathway
single organismal cell-cell adhesion
adherens junction organization
negative regulation of canonical Wnt signaling pathway
receptor binding
protein binding
protein kinase binding
cadherin binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0