Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EP300 SIRNA Assays ›
Gene Symbol:
EP300
Gene Name:
E1A binding protein p300
Gene Aliases:
KAT3B, RSTS2, p300
Chromosome Location:
Chr.22: 41092610 - 41180077 on Build GRCh38
UniGene ID:
Hs.517517
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
145895
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EP300
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001429.3
NP_001420.2
2
526
XM_006724165.3
XP_006724228.1
2
226
GenBank mRNA
BC053889.1
-
2
494
U01877.1
-
2
1330
Back To Top
More Information
Panther Classification:
Molecular Function -
histone modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
response to hypoxia
somitogenesis
liver development
positive regulation of protein phosphorylation
stimulatory C-type lectin receptor signaling pathway
transcription-coupled nucleotide-excision repair
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
protein acetylation
internal protein amino acid acetylation
apoptotic process
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
positive regulation of transcription from RNA polymerase II promoter involved in unfolded protein response
Notch signaling pathway
nervous system development
heart development
skeletal muscle tissue development
memory
circadian rhythm
organ morphogenesis
regulation of autophagy
positive regulation of glycoprotein biosynthetic process
macrophage derived foam cell differentiation
positive regulation of cell death
positive regulation of muscle atrophy
viral process
N-terminal peptidyl-lysine acetylation
internal peptidyl-lysine acetylation
B cell differentiation
platelet formation
lung development
response to cobalt ion
positive regulation of protein binding
positive regulation of type I interferon production
positive regulation of collagen biosynthetic process
positive regulation of protein import into nucleus, translocation
response to tumor necrosis factor
cellular response to UV
positive regulation of histone acetylation
cellular response to drug
megakaryocyte development
cellular response to trichostatin A
intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of DNA binding
protein kinase B signaling
response to estrogen
positive regulation by host of viral transcription
histone H3 acetylation
histone H4 acetylation
histone H2B acetylation
fat cell differentiation
response to ethanol
positive regulation of translation
positive regulation of axon extension
positive regulation of cell size
positive regulation of gene expression, epigenetic
positive regulation of proteolysis
positive regulation of transcription from RNA polymerase II promoter
digestive tract development
positive regulation of protein secretion
protein stabilization
positive regulation of sequence-specific DNA binding transcription factor activity
response to calcium ion
regulation of cell cycle
regulation of angiotensin metabolic process
positive regulation of sarcomere organization
regulation of androgen receptor signaling pathway
regulation of transcription from RNA polymerase II promoter in response to hypoxia
protein-DNA complex assembly
cellular response to hydrogen peroxide
response to fatty acid
cellular response to retinoic acid
cellular response to cAMP
cellular response to glucose stimulus
cellular response to mineralocorticoid stimulus
cellular response to dexamethasone stimulus
regulation of tubulin deacetylation
regulation of cellular response to heat
regulation of signal transduction by p53 class mediator
beta-catenin-TCF complex assembly
cellular response to nerve growth factor stimulus
negative regulation of miRNA metabolic process
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II core promoter sequence-specific DNA binding
core promoter binding
RNA polymerase II activating transcription factor binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
p53 binding
DNA binding
chromatin binding
damaged DNA binding
transcription coactivator activity
histone acetyltransferase activity
lysine N-acetyltransferase activity, acting on acetyl phosphate as donor
protein binding
beta-catenin binding
protein C-terminus binding
transcription factor binding
zinc ion binding
acetyltransferase activity
transferase activity, transferring acyl groups
chromatin DNA binding
protein complex binding
activating transcription factor binding
peptide N-acetyltransferase activity
nuclear hormone receptor binding
glucocorticoid receptor binding
peroxisome proliferator activated receptor binding
bHLH transcription factor binding
SMAD binding
androgen receptor binding
mitogen-activated protein kinase binding
NF-kappaB binding
pre-mRNA intronic binding
protein antigen binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0